missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Erbium foil, 0.025mm (0.001 in.) thick, 99.9% (REO), Thermo Scientific Chemicals

$319.58 - $1035.44

Chemical Identifiers

CAS 7440-52-0
Molecular Formula Er
Molecular Weight (g/mol) 167.26
MDL Number MFCD00010987
InChI Key UYAHIZSMUZPPFV-UHFFFAOYSA-N
Synonym unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic
PubChem CID 23980
ChEBI CHEBI:33379
IUPAC Name erbium
SMILES [Er]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA12388FF
View Documents
Thermo Scientific Chemicals
012388FF
25 x 25 mm
Each for $319.58
 
AA12388FI
View Documents
Thermo Scientific Chemicals
12388FI
50 x 50 mm
Each for $1,035.44
 
Description

Description

Erbium foil used in Physical Vapor Deposition (PVD) processes, including thermal and electron-beam (e-beam) evaporation, for the preparation of thin films. It is used as an alloying element with titanium and is also used to produce infra-red absorbing glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Chemical Identifiers

7440-52-0
167.26
UYAHIZSMUZPPFV-UHFFFAOYSA-N
23980
erbium
Er
MFCD00010987
unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic
CHEBI:33379
[Er]

Specifications

7440-52-0
MFCD00010987
25 x 25 mm
Insoluble in water.
[Er]
167.26
CHEBI:33379
≈0.14g/25x25mm
Erbium foil, 0.025mm (0.001 in.) thick
Er
UN3178
unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic
UYAHIZSMUZPPFV-UHFFFAOYSA-N
erbium
23980
99.9%
Foil
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

EINECSNumber : 231-160-1

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only