missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Erbium foil, 0.62mm (0.024in) thick, 99.9% (REO), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 000593FF
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Erbium foil | |
7440-52-0 | |
Er | |
25 x 25 mm | |
UYAHIZSMUZPPFV-UHFFFAOYSA-N | |
erbium | |
23980 | |
99.9% | |
≈3.5g/25x25mm |
0.62mm (0.024 in.) thick | |
(REO) | |
MFCD00010987 | |
unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic | |
[Er] | |
167.26 | |
CHEBI:33379 | |
Odorless | |
Foil |
Chemical Identifiers
7440-52-0 | |
167.26 | |
UYAHIZSMUZPPFV-UHFFFAOYSA-N | |
23980 | |
erbium |
Er | |
MFCD00010987 | |
unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic | |
CHEBI:33379 | |
[Er] |
Safety and Handling
P210-P240-P241-P280-P370+P378q
H228
EINECSNumber : 231-160-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only