Learn More
Erbium(III) oxide, 99.8% (metals basis), Thermo Scientific Chemicals
For vacuum deposition
$982.90 - $982.90
Chemical Identifiers
CAS | 12061-16-4 |
---|---|
Molecular Formula | Er2O3 |
Molecular Weight (g/mol) | 382.52 |
MDL Number | MFCD00010991 |
InChI Key | ZXGIFJXRQHZCGJ-UHFFFAOYSA-N |
IUPAC Name | dierbium(3+) trioxidandiide |
SMILES | [O--].[O--].[O--].[Er+3].[Er+3] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA3637222
|
Thermo Scientific Chemicals
03637222 |
100 g |
Each for $982.90
|
|
|||||
Description
Erbium(III) oxide is used for vacuum deposition electrical, optical and photoluminescenc, gate dielectrics in semi conductor devices, and in Biomedicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12061-16-4 | |
382.52 | |
ZXGIFJXRQHZCGJ-UHFFFAOYSA-N | |
[O--].[O--].[O--].[Er+3].[Er+3] |
Er2O3 | |
MFCD00010991 | |
dierbium(3+) trioxidandiide |
Specifications
2400°C | |
Sintered Lump | |
Er2O3 | |
14,3646 | |
ZXGIFJXRQHZCGJ-UHFFFAOYSA-N | |
dierbium(3+) trioxidandiide | |
382.56 | |
Hygroscopic | |
Erbium(III) oxide |
12061-16-4 | |
100 g | |
MFCD00010991 | |
Soluble in water(0.0049g/L). | |
[O--].[O--].[O--].[Er+3].[Er+3] | |
382.52 | |
99.8% | |
8.64 g/mL | |
(metals basis) |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Warning
EINECSNumber : 235-045-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only