Learn More
Erbium(III) perchlorate hexahydrate, 50% w/w in aq. soln., Reagent Grade, Thermo Scientific Chemicals
$96.96 - $1197.97
Chemical Identifiers
CAS | 14692-15-0 |
---|---|
Molecular Formula | Cl3ErH12O18 |
Molecular Weight (g/mol) | 573.687 |
MDL Number | MFCD00150232 |
InChI Key | JZRNHNTYRRYOCR-UHFFFAOYSA-K |
Synonym | erbium iii perchlorate hexahydrate, erbium perchlorate-water 1/3/6, erbium iii perchlorate hexahydrate w/w in aqueous solution, reagent grade |
PubChem CID | 20038200 |
IUPAC Name | erbium(3+);triperchlorate;hexahydrate |
SMILES | O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Er+3] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AA40576NA
|
Thermo Scientific Chemicals
040576NA |
∼2 g |
Each for $96.96
|
|
||||||
AA40576NC
|
Thermo Scientific Chemicals
040576NC |
10 g |
Each for $310.45
|
|
||||||
AA40576NE
|
Thermo Scientific Chemicals
040576NE |
∼50 g |
Each for $1,197.97
|
|
||||||
Description
Erbium(III) perchlorate hexahydrate is used in nuclear technology for neutron-absorbing control rods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14692-15-0 | |
573.687 | |
JZRNHNTYRRYOCR-UHFFFAOYSA-K | |
20038200 | |
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Er+3] |
Cl3ErH12O18 | |
MFCD00150232 | |
erbium iii perchlorate hexahydrate, erbium perchlorate-water 1/3/6, erbium iii perchlorate hexahydrate w/w in aqueous solution, reagent grade | |
erbium(3+);triperchlorate;hexahydrate |
Specifications
Pink | |
Liquid | |
Cl3ErH12O18 | |
erbium iii perchlorate hexahydrate, erbium perchlorate-water 1/3/6, erbium iii perchlorate hexahydrate w/w in aqueous solution, reagent grade | |
JZRNHNTYRRYOCR-UHFFFAOYSA-K | |
erbium(3+);triperchlorate;hexahydrate | |
20038200 | |
Reagent | |
Pink liquid |
14692-15-0 | |
∼2 g | |
MFCD00150232 | |
Micible with water. | |
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Er+3] | |
573.687 | |
573.70 (465.61 Anhydrous) | |
Odorless | |
Erbium(III) perchlorate hexahydrate |
Safety and Handling
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H271-H315-H319-H335
DOTInformation : DOT Class: 5.1, Packing Group: II
EINECSNumber : 237-840-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only