Learn More
Erbium(III) sulfate octahydrate, 99.9% (REO), Thermo Scientific Chemicals
$83.36 - $428.27
Chemical Identifiers
CAS | 10031-52-4 |
---|---|
MDL Number | MFCD00149696 |
Synonym | erbium sulfate octahydrate, dierbium trisulfate octahydrate, erbium sulfate er2 so4 3 octahydrate, erbium sulfate hydrate er2 so4 3.8h2o, sulfuric acid, erbium 3+ salt 3:2 , octahydrate, erbium sulfate, erbium sulfate-water 2/3/8, erbium iii sulfate octahydrate reo, dierbium 3+ ion octahydrate trisulfate |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA4009206
|
Thermo Scientific Chemicals
04009206 |
5 g |
Each for $83.36
|
|
|||||
AA4009209
|
Thermo Scientific Chemicals
04009209 |
10 g |
Each for $113.85
|
|
|||||
AA4009218
|
Thermo Scientific Chemicals
04009218 |
50 g |
Each for $428.27
|
|
|||||
Description
Erbium(III) sulfate octahydrate is used as a colorant in glass manufacturing and porcelain enamel glazes. High purity Erbium Sulfate is applied as dopant in making optical fiber.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10031-52-4 | |
MFCD00149696 | |
erbium sulfate octahydrate, dierbium trisulfate octahydrate, erbium sulfate er2 so4 3 octahydrate, erbium sulfate hydrate er2 so4 3.8h2o, sulfuric acid, erbium 3+ salt 3:2 , octahydrate, erbium sulfate, erbium sulfate-water 2/3/8, erbium iii sulfate octahydrate reo, dierbium 3+ ion octahydrate trisulfate |
Specifications
(decomposition) | |
Crystalline | |
MFCD00149696 | |
Soluble in water. | |
99.9% | |
Hygroscopic | |
Erbium(III) sulfate octahydrate |
10031-52-4 | |
5 g | |
erbium sulfate octahydrate, dierbium trisulfate octahydrate, erbium sulfate er2 so4 3 octahydrate, erbium sulfate hydrate er2 so4 3.8h2o, sulfuric acid, erbium 3+ salt 3:2 , octahydrate, erbium sulfate, erbium sulfate-water 2/3/8, erbium iii sulfate octahydrate reo, dierbium 3+ ion octahydrate trisulfate | |
766.82 (622.70 Anhydrous) | |
Odorless | |
3.205 g/mL | |
(REO) |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 236-779-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only