missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Erbium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals
Form: Approx. 0.29g/mm, Cast rod, diameter is approximate
$305.78 - $817.64
Chemical Identifiers
CAS | 7440-52-0 |
---|---|
Molecular Formula | Er |
Molecular Weight (g/mol) | 167.26 |
MDL Number | MFCD00010987 |
InChI Key | UYAHIZSMUZPPFV-UHFFFAOYSA-N |
Synonym | unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic |
PubChem CID | 23980 |
ChEBI | CHEBI:33379 |
IUPAC Name | erbium |
SMILES | [Er] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA40287HF
|
Thermo Scientific Chemicals
040287HF |
10 mm |
Each for $305.78
|
|
|||||
AA40287DH
|
Thermo Scientific Chemicals
040287DH |
25 mm |
Each for $494.10
|
|
|||||
AA40287DJ
|
Thermo Scientific Chemicals
040287DJ |
50 mm |
Each for $817.64
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-52-0 | |
167.26 | |
UYAHIZSMUZPPFV-UHFFFAOYSA-N | |
23980 | |
erbium |
Er | |
MFCD00010987 | |
unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic | |
CHEBI:33379 | |
[Er] |
Specifications
Silver to Gray | |
Cast Rod | |
99.9% (Metals basis excluding Ta) | |
MFCD00010987 | |
UYAHIZSMUZPPFV-UHFFFAOYSA-N | |
erbium | |
23980 | |
Odorless | |
Erbium |
7440-52-0 | |
10 mm | |
Er | |
unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic | |
[Er] | |
167.26 | |
CHEBI:33379 | |
Silver Gray rod |
Safety and Handling
P210-P240-P241-P280-P370+P378q
H228
EINECSNumber : 231-160-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only