Ergodyne™ ProFlex™ 1500 Weight Lifters Style Back Support Brace
Supplier:  Ergodyne™ 11474
 View more versions of this product
Catalog No. 19050804
Explore available promotions
Description
Description
Specifications
Specifications
Black
Proflex™ Back Support
X-Large
Lumbar pad insert
38 to 42 in.
97 to 107 cm
19.7 cm
Adjustable
Contoured
Ideal for over-head lifting, material handling, construction, warehousing, distribution, maintenance, freight/ Baggage, assembly/fabrication, landscaping/grounds
Nylon
1500
7.75 in.