missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ergodyne™ ProFlex™ 4000/4010 Wrist Supports
Flexible support that doesn't compromise productivity
Supplier: Ergodyne™ 70038
Description
- Ideal for material handling work
- Durable supports encourage ergonomically neutral posture
- Limit wrist flexion without compromising finger dexterity
- Patented Open-Center Stay™ reduces pressure on median nerve
- Fully adjustable for an exact fit and comfortable wear
- Exclusive red line indicator ensures proper fit every time
- Anti-odor treatment pulls moisture away from skin
- Machine washable
ProFlex 4000
- Limits downward wrist movement
- Adjustable double-closure stay that limits both upward and downward wrist movements
Order Info
Sold individually. Other styles available. See www.fishersafety.com.
1 Measure around the smallest part of the wrist. This is a general guide only. Call your Fisher Safety Customer Service Representative at 1-800-772-6733 for more information.
Specifications
Black | |
Proflex™ Single Strap Wrist Support | |
Food processing, picking, packing, assembly, fabrication and taffy pulling | |
Left | |
Elastic | |
8 in. |
Two stage hook and loop | |
X-Large | |
Patented open-center Stay™, red line indicator, anti-bacterial lining | |
Limits both upward and downward wrist movements | |
4010 | |
20 cm |