Learn More
Thermo Scientific Chemicals Eriochrome™ Black T, pure, indicator grade
$50.12 - $309.42
Chemical Identifiers
CAS | 1787-61-7 |
---|---|
Molecular Formula | C20H12N3NaO7S |
Molecular Weight (g/mol) | 461.38 |
MDL Number | MFCD00003935 |
InChI Key | JHUJLRKQZAPSDP-GXTSIBQPSA-M |
Synonym | C.I. 14645, 3-Hydroxy-4-(1-hydroxy-2-naphthylazo)-7-nitro-1-naphthalene sulfonic acid, sodium salt, Mordant Black 11 |
PubChem CID | 87355429 |
IUPAC Name | (4Z)-4-[(1-hydroxynaphthalen-2-yl)hydrazinylidene]-7-nitro-3-oxonaphthalene-1-sulfonic acid;sodium |
SMILES | [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC(=CC=C12)[N+]([O-])=O)S([O-])(=O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC228360250
|
Thermo Scientific Chemicals
228360250 |
25 g | Glass bottle |
Each for $50.12
|
|
||||
AC228361000
|
Thermo Scientific Chemicals
228361000 |
100 g | Glass bottle |
Each for $57.41
|
|
||||
AC228365000
|
Thermo Scientific Chemicals
228365000 |
500 g | Glass bottle |
Each for $224.39
|
|
||||
AC228360010
|
Thermo Scientific Chemicals
228360010 |
1 kg | Glass bottle |
Each for $309.42
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
1787-61-7 | |
461.38 | |
JHUJLRKQZAPSDP-GXTSIBQPSA-M | |
87355429 | |
[Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC(=CC=C12)[N+]([O-])=O)S([O-])(=O)=O |
C20H12N3NaO7S | |
MFCD00003935 | |
C.I. 14645, 3-Hydroxy-4-(1-hydroxy-2-naphthylazo)-7-nitro-1-naphthalene sulfonic acid, sodium salt, Mordant Black 11 | |
(4Z)-4-[(1-hydroxynaphthalen-2-yl)hydrazinylidene]-7-nitro-3-oxonaphthalene-1-sulfonic acid;sodium |
Specifications
1787-61-7 | |
MFCD00003935 | |
15, 3725 | |
Solubility in water: slightly soluble. | |
Authentic | |
(4Z)-4-[(1-hydroxynaphthalen-2-yl)hydrazinylidene]-7-nitro-3-oxonaphthalene-1-sulfonic acid;sodium | |
87355429 | |
Pure | |
Passes (for complexometric determination of metals) | |
Black to Black/Brown | |
Powder |
C20H12N3NaO7S | |
16, III, 320 | |
C.I. 14645, 3-Hydroxy-4-(1-hydroxy-2-naphthylazo)-7-nitro-1-naphthalene sulfonic acid, sodium salt, Mordant Black 11 | |
JHUJLRKQZAPSDP-GXTSIBQPSA-M | |
[Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC(=CC=C12)[N+]([O-])=O)S([O-])(=O)=O | |
461.38 | |
461.38 | |
Glass bottle | |
Complies | |
25 g | |
Eriochrome™ Black T |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 217-250-3
RTECSNumber : QK2197000
TSCA : TSCA
RUO – Research Use Only