missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eriochrome™ Black T Solution, Fisher Chemical
Supplier: Fisher Chemical SE32500
Description
EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.
Specifications
Eriochrome™ Black T Solution | |
64°C | |
CH4O | |
Mordant Black 11 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
Glass Bottle | |
97mmHg | |
Blue | |
Liquid |
-98°C | |
1787-61-7,1336-21-6,7732-18-5 | |
MFCD00004595 | |
Pass Test | |
CO | |
32.04 | |
Pass Test | |
14645 | |
500 mL |