Greiner Bio-One Erlenmeyer Shaker Flasks with Baffled Bottom
Ideal for suspension culture of mammalian cells, plants, microbes and microorganisms. Each flask is manufactured utilizing an optically clear, inert polycarbonate resin which can be disposed of after single use or be made reusable when autoclaved.
Supplier:  Greiner Bio-One 679516
 View more versions of this product
Catalog No. 07-001-075
Explore available promotions
Description
Description
  • 2-in-1 DuoCAP™ for flexibility of sterile air exchange
    or tight seal
  • Made of optically clear polycarbonate which is
    lighter and safer than glass
  • Individually packaged and sterile to SAL 10-6 for
    long term culture applications
  • Molded-in graduations for easy volumetric reference
  • Non-pyrogenic, non-cytotoxic, and nuclease-free
  • Available with flat or baffled base for increased
    agitation and aeration
Specifications
Specifications
Clear
Polycarbonate
260 mm (w/cap)
Sterile
3000 mL
Suspension culture
230.5 mm base
Baffled