missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Erucic acid, tech. 90%, Thermo Scientific Chemicals

$69.66 - $231.07

Chemical Identifiers

CAS 112-86-7
Molecular Formula C22H42O2
Molecular Weight (g/mol) 338.576
MDL Number MFCD00063188
InChI Key DPUOLQHDNGRHBS-KTKRTIGZSA-N
Synonym erucic acid, cis-13-docosenoic acid, z-docos-13-enoic acid, 13-docosenoic acid, z, cis-erucic acid, 13-cis-docosenoic acid, 13-docosenoic acid, z-13-docosenoic acid, hystrene 2290
PubChem CID 5281116
ChEBI CHEBI:28792
IUPAC Name (Z)-docos-13-enoic acid
SMILES CCCCCCCCC=CCCCCCCCCCCCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0868518
View Documents
Thermo Scientific Chemicals
L0868518
50 g
Each for $69.66
 
AAL0868530
View Documents
Thermo Scientific Chemicals
L0868530
250 g
Each for $231.07
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

112-86-7
338.576
DPUOLQHDNGRHBS-KTKRTIGZSA-N
5281116
(Z)-docos-13-enoic acid
C22H42O2
MFCD00063188
erucic acid, cis-13-docosenoic acid, z-docos-13-enoic acid, 13-docosenoic acid, z, cis-erucic acid, 13-cis-docosenoic acid, 13-docosenoic acid, z-13-docosenoic acid, hystrene 2290
CHEBI:28792
CCCCCCCCC=CCCCCCCCCCCCC(=O)O

Specifications

112-86-7
265°C (15 mmHg)
C22H42O2
50 g
14,3674
DPUOLQHDNGRHBS-KTKRTIGZSA-N
(Z)-docos-13-enoic acid
5281116
338.58
Technical
29°C to 33°C
>110°C (230°F)
MFCD00063188
1728049
erucic acid, cis-13-docosenoic acid, z-docos-13-enoic acid, 13-docosenoic acid, z, cis-erucic acid, 13-cis-docosenoic acid, 13-docosenoic acid, z-13-docosenoic acid, hystrene 2290
CCCCCCCCC=CCCCCCCCCCCCC(=O)O
338.576
CHEBI:28792
90%
Erucic acid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 204-011-3

RTECSNumber : JR1280000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only