missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Erythrosin B, Thermo Scientific Chemicals

Erythrosin B, C40H14I8Na2O10, CAS Number-16423-68-0, C.I. 45430; Tetraiodofluorescein sodium salt, 500g, 240-046-0, 879.87, H302, GHS H Statement H302 Harmful if swallowed., P264b-P270-P301+P312-P330-P501c, DKRYETBOLGDIOK-UHFFFAOYSA-L

Supplier:  Thermo Scientific Chemicals A1418022

 View more versions of this product

Catalog No. AAA1418022


Explore available promotions

Description

Description

Erythrosin B, also called Acid Red 51, is used as a food colorant, in printing inks and as a biological stain in plant anatomy. It finds application as a dental plaque disclosing agent and a sensitizer for orthochromatic photographic films. As a food additive, it is commonly used in sweets like candies, popsicles and in cake-decorating gels. It is an important dye used in the determination of cell viability and used as phosphorescent triplet probe to detect rotational diffusion of membrane proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Erythrosin B, also called Acid Red 51, is used as a food colorant, in printing inks and as a biological stain in plant anatomy. It finds application as a dental plaque disclosing agent and a sensitizer for orthochromatic photographic films. As a food additive, it is commonly used in sweets like candies, popsicles and in cake-decorating gels. It is an important dye used in the determination of cell viability and used as phosphorescent triplet probe to detect rotational diffusion of membrane proteins.

Solubility
Soluble in water, glycerin and methanol.

Notes
Light sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place.Incompatible with oxidizing agents.
Specifications

Specifications

Erythrosin B
C40H14I8Na2O10
C.I. 45430; Tetraiodofluorescein sodium salt
DKRYETBOLGDIOK-UHFFFAOYSA-L
disodium;2-(3-hydroxy-2,4,5,7-tetraiodo-6-oxoxanthen-9-yl)benzoic acid;2-(2,4,5,7-tetraiodo-3-oxido-6-oxoxanthen-9-yl)benzoate
131676154
Hygroscopic; Light sensitive
16423-68-0
MFCD00144257
Soluble in water,glycerin and methanol.
C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)I)O)I)I)I)C(=O)O.C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)I)[O-])I)I)I)C(=O)[O-].[Na+].[Na+]
1715.757
879.87
100 g

Chemical Identifiers

16423-68-0
MFCD00144257
C.I. 45430; Tetraiodofluorescein sodium salt
131676154
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 240-046-0

RTECSNumber : LM5950000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only