missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Estra-5(10),9(11)-diene-3,17-dione 3-Ethylene Ketal 98.0+%, TCI America™
Supplier: TCI America E09181G
Specifications
Estra-5(10),9(11)-diene-3,17-dione 3-Ethylene Ketal | |
156°C | |
C20H26O3 | |
1 g | |
XUOQKQRMICQUQC-AOIWGVFYSA-N | |
(8S,13S,14S)-13-methylspiro[1,2,4,6,7,8,12,14,15,16-decahydrocyclopenta[a]phenanthrene-3,2'-1,3-dioxolane]-17-one | |
7076399 | |
≥98.0% (HPLC) |
5571-36-8 | |
White-Yellow | |
MFCD08062585 | |
3077 | |
CC12CC=C3C(C1CCC2=O)CCC4=C3CCC5(C4)OCCO5 | |
314.425 | |
314.43 | |
Crystalline Powder |
Chemical Identifiers
5571-36-8 | |
314.425 | |
XUOQKQRMICQUQC-AOIWGVFYSA-N | |
(8S,13S,14S)-13-methylspiro[1,2,4,6,7,8,12,14,15,16-decahydrocyclopenta[a]phenanthrene-3,2'-1,3-dioxolane]-17-one |
C20H26O3 | |
MFCD08062585 | |
7076399 | |
CC12CC=C3C(C1CCC2=O)CCC4=C3CCC5(C4)OCCO5 |
Safety and Handling
TSCA : No