missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Estra-5(10),9(11)-diene-3,17-dione 3-Ethylene Ketal 98.0+%, TCI America™

Supplier:  TCI America E09185G

 View more versions of this product

Catalog No. E09185G


Explore available promotions

Description

Description

Specifications

Specifications

Estra-5(10),9(11)-diene-3,17-dione 3-Ethylene Ketal
156°C
C20H26O3
5 g
XUOQKQRMICQUQC-AOIWGVFYSA-N
(8S,13S,14S)-13-methylspiro[1,2,4,6,7,8,12,14,15,16-decahydrocyclopenta[a]phenanthrene-3,2'-1,3-dioxolane]-17-one
7076399
≥98.0% (HPLC)
5571-36-8
White-Yellow
MFCD08062585
3077
CC12CC=C3C(C1CCC2=O)CCC4=C3CCC5(C4)OCCO5
314.425
314.43
Crystalline Powder

Chemical Identifiers

5571-36-8
314.425
XUOQKQRMICQUQC-AOIWGVFYSA-N
(8S,13S,14S)-13-methylspiro[1,2,4,6,7,8,12,14,15,16-decahydrocyclopenta[a]phenanthrene-3,2'-1,3-dioxolane]-17-one
C20H26O3
MFCD08062585
7076399
CC12CC=C3C(C1CCC2=O)CCC4=C3CCC5(C4)OCCO5
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions