missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Estramustine phosphate sodium

$468.26 - $1388.58

Chemical Identifiers

CAS 52205-73-9
Molecular Formula C23H30Cl2NNa2O6P
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC466740010
View Documents
Thermo Scientific Chemicals
466740010
1 g
Each for $1,388.58
Only null left
 
AC466742500
View Documents
Thermo Scientific Chemicals
466742500
250 mg
Each for $468.26
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

52205-73-9
C23H30Cl2NNa2O6P

Specifications

52205-73-9
Solubility aq. soln.: 50 g/L
564.35
[Na+].[Na+].C[C@]12CC[C@H]3[C@@H](CCC4=CC(OC(=O)N(CCCl)CCCl)=CC=C34)[C@@H]1CC[C@@H]2OP([O-])([O-])=O
564.35
1 g
C23H30Cl2NNa2O6P
IIUMCNJTGSMNRO-VVSKJQCTSA-L
disodium (1S,3aS,3bR,9bS,11aS)-7-{[bis(2-chloroethyl)carbamoyl]oxy}-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl phosphate
Estramustine phosphate sodium
Videos
Safety and Handling

Safety and Handling

GHS H Statement Causes skin irritation.

May cause an allergic skin reaction.

Causes serious eye damage.

May cause respiratory irritation.

Suspected of damaging fertility or the unborn child.

GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Immediately call a POISON CENTER or doctor/physician.

Wear protective gloves/protective clothing/eye protection/face protection.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Danger

EINECSNumber : 257-735-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only