Learn More
Ethanesulfonyl chloride, 98+%, Thermo Scientific Chemicals
$52.61 - $52.61
Chemical Identifiers
CAS | 594-44-5 |
---|---|
Molecular Formula | C2H5ClO2S |
Molecular Weight (g/mol) | 128.57 |
MDL Number | MFCD00007460 |
InChI Key | FRYHCSODNHYDPU-UHFFFAOYSA-N |
Synonym | ethylsulfonyl chloride, ethylsulfochloride, 1-ethanesulfonyl chloride, ethanesulphonyl chloride, ethanesulfonylchloride, unii-b56zoc898a, ethylsulfonylchloride, ethansulfonyl chloride, ethansulfonylchlorid, ethanesulfonylchoride |
PubChem CID | 11667 |
IUPAC Name | ethanesulfonyl chloride |
SMILES | CCS(Cl)(=O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC215820050
|
Thermo Scientific Chemicals
215820050 |
5 mL | Glass bottle |
Each for $52.61
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
594-44-5 | |
128.57 | |
FRYHCSODNHYDPU-UHFFFAOYSA-N | |
11667 | |
CCS(Cl)(=O)=O |
C2H5ClO2S | |
MFCD00007460 | |
ethylsulfonyl chloride, ethylsulfochloride, 1-ethanesulfonyl chloride, ethanesulphonyl chloride, ethanesulfonylchloride, unii-b56zoc898a, ethylsulfonylchloride, ethansulfonyl chloride, ethansulfonylchlorid, ethanesulfonylchoride | |
ethanesulfonyl chloride |
Specifications
594-44-5 | |
1.3500g/mL | |
83°C | |
98% min. (GC) | |
C2H5ClO2S | |
C2H5SO2Cl | |
5 mL | |
1.35 | |
Solubility in water: decomposes in water. Other solubilities: soluble in acetone and ethyl ether | |
CCS(Cl)(=O)=O | |
128.57 | |
1.44 mm2/s (20°C) | |
98+% | |
Ethanesulfonyl chloride, 98+% |
Brown to Pink-Purple or Colorless to Yellow | |
177.0°C | |
Authentic | |
Glass bottle | |
1.4520 to 1.4540 | |
MFCD00007460 | |
04, 6 | |
ethylsulfonyl chloride, ethylsulfochloride, 1-ethanesulfonyl chloride, ethanesulphonyl chloride, ethanesulfonylchloride, unii-b56zoc898a, ethylsulfonylchloride, ethansulfonyl chloride, ethansulfonylchlorid, ethanesulfonylchoride | |
FRYHCSODNHYDPU-UHFFFAOYSA-N | |
ethanesulfonyl chloride | |
11667 | |
128.58 | |
Liquid |
Safety and Handling
GHS H Statement:
Toxic if inhaled.
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 209-842-5
RTECSNumber : KI8050000
TSCA : TSCA
Recommended Storage : this product darkens during storage
RUO – Research Use Only