Learn More
Ethanethiol, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 117865000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethanethiol | |
75-08-1 | |
Colorless | |
35.0°C | |
Authentic | |
Glass bottle | |
1.4296 to 1.4316 | |
MFCD00004887 | |
01,34 | |
0.83 | |
ethyl mercaptan, mercaptoethane, thioethanol, ethyl hydrosulfide, ethyl sulfhydrate, ethyl thioalcohol, thioethyl alcohol, 1-mercaptoethane, aethanethiol, aethylmercaptan | |
DNJIEGIFACGWOD-UHFFFAOYSA-N | |
ethanethiol | |
6343 | |
99+% |
99+% | |
-148.0°C | |
0.8300g/mL | |
−45°C | |
99% min. (GC) | |
C2H6S | |
C2H5SH | |
500 mL | |
12,29 | |
15,3781 | |
Solubility in water: 6.76g/L (20°C). Other solubilities: soluble in alcohol and ether | |
CCS | |
62.13 | |
62.13 | |
Liquid |
Chemical Identifiers
75-08-1 | |
62.13 | |
DNJIEGIFACGWOD-UHFFFAOYSA-N | |
6343 | |
CCS |
C2H6S | |
MFCD00004887 | |
ethyl mercaptan, mercaptoethane, thioethanol, ethyl hydrosulfide, ethyl sulfhydrate, ethyl thioalcohol, thioethyl alcohol, 1-mercaptoethane, aethanethiol, aethylmercaptan | |
ethanethiol |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Very toxic to aquatic life with long lasting effects.
Extremely flammable liquid and vapor.
GHS P Statement
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away
GHS Signal Word: Danger
EINECSNumber : 200-837-3
RTECSNumber : KI9625000
TSCA : TSCA
RUO – Research Use Only