missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethosuximide, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 461400050

 View more versions of this product

Catalog No. AC461400050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Ethosuximide
134.0°C (5.0 mmHg)
C7H11NO2
5g
Solubility in water: soluble.
CCC1(C)CC(=O)NC1=O
141.17
CHEBI:4887
≥97.5% (HPLC)
77-67-8
Authentic
MFCD00072123
ethosuximide, zarontin, etosuximida, 2-ethyl-2-methylsuccinimide, ethosuxide, ethosuccimide, ethosuccinimide, petnidan, suxinutin, atysmal
HAPOVYFOVVWLRS-UHFFFAOYNA-N
3-ethyl-3-methylpyrrolidine-2,5-dione
3291
141.17

Chemical Identifiers

77-67-8
141.17
HAPOVYFOVVWLRS-UHFFFAOYNA-N
3291
3-ethyl-3-methylpyrrolidine-2,5-dione
C7H11NO2
MFCD00072123
ethosuximide, zarontin, etosuximida, 2-ethyl-2-methylsuccinimide, ethosuxide, ethosuccimide, ethosuccinimide, petnidan, suxinutin, atysmal
CHEBI:4887
CCC1(C)CC(=O)NC1=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement Harmful if swallowed. Suspected of causing genetic defects. May damage the unborn child.

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face, hands and any exposed skin thoroughly after handling. Obtain special instructions before use. Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Danger

EINECSNumber : 201-048-7

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only