missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethoxyamine hydrochloride, 98%, Thermo Scientific Chemicals

$481.25 - $953.75

Chemical Identifiers

CAS 3332-29-4
Molecular Formula C2H7NO·HCl
Molecular Weight (g/mol) 97.54
MDL Number MFCD00012956
InChI Key NUXCOKIYARRTDC-UHFFFAOYSA-N
Synonym ethoxyamine hydrochloride, o-ethylhydroxylamine hydrochloride, hydroxylamine, o-ethyl-, hydrochloride, ethoxyamine hcl, hydroxylamine, o-ethyl-, hydrochloride 1:1, ethoxyaminehydrochloride, aminooxy ethane hydrochloride 1:1, etonh2.hcl, ethoxylamine hydrochloride, nh2och2ch3.hcl
PubChem CID 76850
IUPAC Name O-ethylhydroxylamine;hydrochloride
SMILES CCON.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC409610100
SDS View Documents
Thermo Scientific Chemicals
409610100
10 g Glass bottle N/A
 
AC409610250
SDS View Documents
Thermo Scientific Chemicals
409610250
25 g Glass bottle N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3332-29-4
97.54
NUXCOKIYARRTDC-UHFFFAOYSA-N
76850
CCON.Cl
C2H7NO·HCl
MFCD00012956
ethoxyamine hydrochloride, o-ethylhydroxylamine hydrochloride, hydroxylamine, o-ethyl-, hydrochloride, ethoxyamine hcl, hydroxylamine, o-ethyl-, hydrochloride 1:1, ethoxyaminehydrochloride, aminooxy ethane hydrochloride 1:1, etonh2.hcl, ethoxylamine hydrochloride, nh2och2ch3.hcl
O-ethylhydroxylamine;hydrochloride

Specifications

3332-29-4
Gray
99% min. (Argentometry)
C2H7NO·HCl
MFCD00012956
ethoxyamine hydrochloride, o-ethylhydroxylamine hydrochloride, hydroxylamine, o-ethyl-, hydrochloride, ethoxyamine hcl, hydroxylamine, o-ethyl-, hydrochloride 1:1, ethoxyaminehydrochloride, aminooxy ethane hydrochloride 1:1, etonh2.hcl, ethoxylamine hydrochloride, nh2och2ch3.hcl
NUXCOKIYARRTDC-UHFFFAOYSA-N
O-ethylhydroxylamine;hydrochloride
76850
99+%
Ethoxyamine hydrochloride, 99+%
130°C to 133°C
Authentic
Glass bottle
C2H5ONH2·HCl
10 g
Solubility in water: soluble
CCON.Cl
97.54
97.54
Crystals, Powder or Flakes
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

EINECSNumber : 222-060-9

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only