missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(Ethoxycarbonylmethyl)dimethylsulfonium bromide, 98%, Thermo Scientific Chemicals

$116.33 - $387.15

Chemical Identifiers

CAS 5187-82-6
Molecular Formula C6H13BrO2S
Molecular Weight (g/mol) 229.132
MDL Number MFCD00830557
InChI Key JXFPTJYKYKVENJ-UHFFFAOYSA-M
Synonym ethoxycarbonylmethyl dimethylsulfonium bromide, 2-ethoxy-2-oxoethyl dimethylsulfonium bromide, carbethoxymethyl-dimethylsulfonium bromide, 2-ethoxy-2-oxoethyl dimethylsulfanium bromide, 2-ethoxy-2-oxoethyl dimethyl sulfonium bromide, 2-ethoxy-2-oxoethyl dimethylsulphonium bromide, sulfonium, carbethoxymethyl dimethyl-, bromide, sulfonium, 2-ethoxy-2-oxoethyl dimethyl-, bromide, carbethoxymethyl dimethylsulfonium bromide, acmc-1az78
PubChem CID 78861
IUPAC Name (2-ethoxy-2-oxoethyl)-dimethylsulfanium;bromide
SMILES CCOC(=O)C[S+](C)C.[Br-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0026609
View Documents
Thermo Scientific Chemicals
L0026609
10 g
Each for $116.33
 
AAL0026618
View Documents
Thermo Scientific Chemicals
L0026618
50 g
Each for $387.15
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5187-82-6
229.132
JXFPTJYKYKVENJ-UHFFFAOYSA-M
78861
CCOC(=O)C[S+](C)C.[Br-]
C6H13BrO2S
MFCD00830557
ethoxycarbonylmethyl dimethylsulfonium bromide, 2-ethoxy-2-oxoethyl dimethylsulfonium bromide, carbethoxymethyl-dimethylsulfonium bromide, 2-ethoxy-2-oxoethyl dimethylsulfanium bromide, 2-ethoxy-2-oxoethyl dimethyl sulfonium bromide, 2-ethoxy-2-oxoethyl dimethylsulphonium bromide, sulfonium, carbethoxymethyl dimethyl-, bromide, sulfonium, 2-ethoxy-2-oxoethyl dimethyl-, bromide, carbethoxymethyl dimethylsulfonium bromide, acmc-1az78
(2-ethoxy-2-oxoethyl)-dimethylsulfanium;bromide

Specifications

5187-82-6
C6H13BrO2S
10 g
Hygroscopic
JXFPTJYKYKVENJ-UHFFFAOYSA-M
(2-ethoxy-2-oxoethyl)-dimethylsulfanium;bromide
78861
98%
∽80°C (decomposition)
MFCD00830557
4161472
ethoxycarbonylmethyl dimethylsulfonium bromide, 2-ethoxy-2-oxoethyl dimethylsulfonium bromide, carbethoxymethyl-dimethylsulfonium bromide, 2-ethoxy-2-oxoethyl dimethylsulfanium bromide, 2-ethoxy-2-oxoethyl dimethyl sulfonium bromide, 2-ethoxy-2-oxoethyl dimethylsulphonium bromide, sulfonium, carbethoxymethyl dimethyl-, bromide, sulfonium, 2-ethoxy-2-oxoethyl dimethyl-, bromide, carbethoxymethyl dimethylsulfonium bromide, acmc-1az78
CCOC(=O)C[S+](C)C.[Br-]
229.132
229.14
(Ethoxycarbonylmethyl)dimethylsulfonium bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 225-968-3

RTECSNumber : WR7600000

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only