missing translation for 'onlineSavingsMsg'
Learn More

Ethoxytrimethylsilane, 97%, Thermo Scientific Chemicals

$206.53 - $206.53

Chemical Identifiers

CAS 1825-62-3
Molecular Formula C5H14OSi
Molecular Weight (g/mol) 118.25
MDL Number MFCD00009069
InChI Key RSIHJDGMBDPTIM-UHFFFAOYSA-N
Synonym trimethylethoxysilane, silane, ethoxytrimethyl, silane, trimethylethoxy, ethyl trimethylsilyl ether, trimethylsilyl ethyl ether, ethoxy trimethyl silane, trimethyl ethoxy silane, silane,ethoxytrimethyl, acmc-1btam, ethanol, tms derivative
PubChem CID 15772
IUPAC Name ethoxy(trimethyl)silane
SMILES CCO[Si](C)(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1015122
View Documents
Thermo Scientific Chemicals
L1015122
100 g
Each for $206.53
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1825-62-3
118.25
RSIHJDGMBDPTIM-UHFFFAOYSA-N
15772
CCO[Si](C)(C)C
C5H14OSi
MFCD00009069
trimethylethoxysilane, silane, ethoxytrimethyl, silane, trimethylethoxy, ethyl trimethylsilyl ether, trimethylsilyl ethyl ether, ethoxy trimethyl silane, trimethyl ethoxy silane, silane,ethoxytrimethyl, acmc-1btam, ethanol, tms derivative
ethoxy(trimethyl)silane

Specifications

75°C to 76°C
1825-62-3
C5H14OSi
UN1993
−18°C (−0°F)
RSIHJDGMBDPTIM-UHFFFAOYSA-N
ethoxy(trimethyl)silane
15772
97%
1.374
0.757
-83°C
100 g
MFCD00009069
1731950
trimethylethoxysilane, silane, ethoxytrimethyl, silane, trimethylethoxy, ethyl trimethylsilyl ether, trimethylsilyl ethyl ether, ethoxy trimethyl silane, trimethyl ethoxy silane, silane,ethoxytrimethyl, acmc-1btam, ethanol, tms derivative
CCO[Si](C)(C)C
118.25
118.25
Mild
Moisture sensitive
Ethoxytrimethylsilane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 217-370-6

RTECSNumber : VV4110000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only