missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 1,2-Benzisoxazole-3-carboxylate 98.0+%, TCI America™
Supplier: TCI America E0731100MG
Specifications
Ethyl 1,2-Benzisoxazole-3-carboxylate | |
59°C | |
C10H9NO3 | |
100 mg | |
IRTACBSCHHOIPA-UHFFFAOYSA-N | |
ethyl 1,2-benzoxazole-3-carboxylate | |
14177940 | |
≥98.0% (GC) |
57764-49-5 | |
Yellow | |
MFCD06659787 | |
1,2-Benzisoxazole-3-carboxylic Acid Ethyl Ester, 3-Ethoxycarbonyl-1,2-benzisoxazole | |
CCOC(=O)C1=NOC2=CC=CC=C12 | |
191.19 | |
191.19 | |
Crystalline Powder |
Chemical Identifiers
57764-49-5 | |
191.19 | |
IRTACBSCHHOIPA-UHFFFAOYSA-N | |
14177940 | |
CCOC(=O)C1=NOC2=CC=CC=C12 |
C10H9NO3 | |
MFCD06659787 | |
1,2-Benzisoxazole-3-carboxylic Acid Ethyl Ester, 3-Ethoxycarbonyl-1,2-benzisoxazole | |
ethyl 1,2-benzoxazole-3-carboxylate |
Safety and Handling
TSCA : No