missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1,4-Benzodioxane-2-carboxylate 98.0+%, TCI America™

$197.59 - $635.65

Chemical Identifiers

CAS 4739-94-0
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.213
MDL Number MFCD01099774
InChI Key DDIGEMWIKJMEIU-UHFFFAOYSA-N
Synonym 1,4-Benzodioxane-2-carboxylic Acid Ethyl Ester
PubChem CID 151226
IUPAC Name ethyl 2,3-dihydro-1,4-benzodioxine-3-carboxylate
SMILES CCOC(=O)C1COC2=CC=CC=C2O1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
E09025G
View Documents
TCI America
E09025G
5 g
Each for $197.59
Only null left
 
E090225G
View Documents
TCI America
E090225G
25 g
Each for $635.65
Only null left
 
Description

Description

Specifications

Chemical Identifiers

4739-94-0
208.213
DDIGEMWIKJMEIU-UHFFFAOYSA-N
151226
CCOC(=O)C1COC2=CC=CC=C2O1
C11H12O4
MFCD01099774
1,4-Benzodioxane-2-carboxylic Acid Ethyl Ester
ethyl 2,3-dihydro-1,4-benzodioxine-3-carboxylate

Specifications

4739-94-0
115°C
MFCD01099774
1,4-Benzodioxane-2-carboxylic Acid Ethyl Ester
CCOC(=O)C1COC2=CC=CC=C2O1
208.213
208.21
Liquid
Yellow
C11H12O4
5 g
DDIGEMWIKJMEIU-UHFFFAOYSA-N
ethyl 2,3-dihydro-1,4-benzodioxine-3-carboxylate
151226
≥98.0% (GC)
Ethyl 1,4-Benzodioxane-2-carboxylate
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'