missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 1-hydroxycyclopropanecarboxylate, 90%, Thermo Scientific Chemicals

$364.48 - $364.48

Chemical Identifiers

CAS 137682-89-4
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD01863257
InChI Key DPMGQZGXWRHYPE-UHFFFAOYSA-N
Synonym ethyl 1-hydroxycyclopropanecarboxylate, 1-hydroxycyclopropanecarboxylic acid ethyl ester, cyclopropanecarboxylic acid, 1-hydroxy-, ethyl ester, 1-ethoxycarbonyl cyclopropan-1-ol, ethyl-1-hydroxycyclopropane carboxylate, 1-hydroxy-cyclopropanecarboxylic acid ethyl ester
PubChem CID 2733706
IUPAC Name ethyl 1-hydroxycyclopropane-1-carboxylate
SMILES CCOC(=O)C1(O)CC1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC335300010
SDS View Documents
Thermo Scientific Chemicals
335300010
1 g Glass bottle
Each for $364.48
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

137682-89-4
130.14
DPMGQZGXWRHYPE-UHFFFAOYSA-N
2733706
CCOC(=O)C1(O)CC1
C6H10O3
MFCD01863257
ethyl 1-hydroxycyclopropanecarboxylate, 1-hydroxycyclopropanecarboxylic acid ethyl ester, cyclopropanecarboxylic acid, 1-hydroxy-, ethyl ester, 1-ethoxycarbonyl cyclopropan-1-ol, ethyl-1-hydroxycyclopropane carboxylate, 1-hydroxy-cyclopropanecarboxylic acid ethyl ester
ethyl 1-hydroxycyclopropane-1-carboxylate

Specifications

137682-89-4
78.0°C (15.0 mmHg)
88% min. (GC)
C6H10O3
1.4400 to 1.4470
1 g
Solubility in water: .. Other solubilities: soluble in most common organic solvents
CCOC(=O)C1(O)CC1
130.14
130.14
Liquid
Yellow to Brown
Authentic
Glass bottle
C3H4(OH)COOCH2CH3
MFCD01863257
ethyl 1-hydroxycyclopropanecarboxylate, 1-hydroxycyclopropanecarboxylic acid ethyl ester, cyclopropanecarboxylic acid, 1-hydroxy-, ethyl ester, 1-ethoxycarbonyl cyclopropan-1-ol, ethyl-1-hydroxycyclopropane carboxylate, 1-hydroxy-cyclopropanecarboxylic acid ethyl ester
DPMGQZGXWRHYPE-UHFFFAOYSA-N
ethyl 1-hydroxycyclopropane-1-carboxylate
2733706
90%
Ethyl 1-hydroxycyclopropanecarboxylate, 90%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only