missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 1-(hydroxymethyl)cyclobutanecarboxylate, 97%, Thermo Scientific Chemicals
$124.01 - $1577.82
Chemical Identifiers
CAS | 1195-81-9 |
---|---|
Molecular Formula | C8H14O3 |
Molecular Weight (g/mol) | 158.197 |
MDL Number | MFCD11707219 |
InChI Key | PWMQFMMZBJUHID-UHFFFAOYSA-N |
Synonym | ethyl 1-hydroxymethyl cyclobutane-1-carboxylate, ethyl 1-hydroxymethylcyclobutanecarboxylate, ethyl 1-hydroxymethyl cyclobutanecarboxylate, cyclobutanecarboxylic acid, 1-hydroxymethyl-, ethyl ester, ethyl 1-hydroxymethyl-cyclobutanecarboxylate, 1-hydroxymethyl-cyclobutanecarboxylic acid ethyl ester |
PubChem CID | 15670506 |
IUPAC Name | ethyl 1-(hydroxymethyl)cyclobutane-1-carboxylate |
SMILES | CCOC(=O)C1(CCC1)CO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH62566MD
|
Thermo Scientific Chemicals
H62566MD |
250 mg |
Each for $124.01
|
|
||||||
AAH6256603
|
Thermo Scientific Chemicals
H6256603 |
1 g |
Each for $350.63
|
|
||||||
AAH6256606
|
Thermo Scientific Chemicals
H6256606 |
5 g |
Each for $1,577.82
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1195-81-9 | |
158.197 | |
PWMQFMMZBJUHID-UHFFFAOYSA-N | |
15670506 | |
CCOC(=O)C1(CCC1)CO |
C8H14O3 | |
MFCD11707219 | |
ethyl 1-hydroxymethyl cyclobutane-1-carboxylate, ethyl 1-hydroxymethylcyclobutanecarboxylate, ethyl 1-hydroxymethyl cyclobutanecarboxylate, cyclobutanecarboxylic acid, 1-hydroxymethyl-, ethyl ester, ethyl 1-hydroxymethyl-cyclobutanecarboxylate, 1-hydroxymethyl-cyclobutanecarboxylic acid ethyl ester | |
ethyl 1-(hydroxymethyl)cyclobutane-1-carboxylate |
Specifications
1195-81-9 | |
MFCD11707219 | |
ethyl 1-hydroxymethyl cyclobutane-1-carboxylate, ethyl 1-hydroxymethylcyclobutanecarboxylate, ethyl 1-hydroxymethyl cyclobutanecarboxylate, cyclobutanecarboxylic acid, 1-hydroxymethyl-, ethyl ester, ethyl 1-hydroxymethyl-cyclobutanecarboxylate, 1-hydroxymethyl-cyclobutanecarboxylic acid ethyl ester | |
CCOC(=O)C1(CCC1)CO | |
158.197 | |
158.19 | |
Ethyl 1-(hydroxymethyl)cyclobutanecarboxylate |
C8H14O3 | |
250 mg | |
PWMQFMMZBJUHID-UHFFFAOYSA-N | |
ethyl 1-(hydroxymethyl)cyclobutane-1-carboxylate | |
15670506 | |
97% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only