missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1H-pyrazole-4-carboxylate, 98%

CAS: 37622-90-5 | C6H8N2O2 | 140.142 g/mol

$263.03 - $1045.05

Chemical Identifiers

CAS 37622-90-5
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.142
MDL Number MFCD00010844
InChI Key KACZQOKEFKFNDB-UHFFFAOYSA-N
PubChem CID 142179
IUPAC Name ethyl 1H-pyrazole-4-carboxylate
SMILES CCOC(=O)C1=CNN=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2535106
View Documents
Thermo Scientific Chemicals
B2535106
5 g
Each for $263.03
Only null left
 
AAB2535114
View Documents
Thermo Scientific Chemicals
B2535114
25 g
Each for $1,045.05
Only null left
 
Description

Description

Ethyl 1H-pyrazole-4-carboxylate is used in the preparation of isoxazole-4-carboxylic acid derivatives and isoxazole-3,5-dicarboxamides. Further, it acts as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 1H-pyrazole-4-carboxylate is used in the preparation of isoxazole-4-carboxylic acid derivatives and isoxazole-3,5-dicarboxamides. Further, it acts as an intermediate in organic synthesis.

Solubility
Miscible with acetone.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.
Specifications

Chemical Identifiers

37622-90-5
140.142
KACZQOKEFKFNDB-UHFFFAOYSA-N
ethyl 1H-pyrazole-4-carboxylate
C6H8N2O2
MFCD00010844
142179
CCOC(=O)C1=CNN=C1

Specifications

37622-90-5
138°C to 140°C (3 mmHg)
MFCD00010844
ethyl 4-pyrazolecarboxylate, ethyl pyrazole-4-carboxylate, 1h-pyrazole-4-carboxylic acid ethyl ester, 4-ethoxycarbonylpyrazole, 1h-pyrazole-4-carboxylic acid, ethyl ester, 4-pyrazolecarboxylic acid ethyl ester, ethyl4-pyrazolecarboxylate, zlchem 132, pubchem21939, acmc-209iug
KACZQOKEFKFNDB-UHFFFAOYSA-N
ethyl 1H-pyrazole-4-carboxylate
142179
98%
75°C to 80°C
C6H8N2O2
5 g
Miscible with acetone.
CCOC(=O)C1=CNN=C1
140.142
140.14
Ethyl 1H-pyrazole-4-carboxylate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only