missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2-chloro-2-(hydroxyimino)acetate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 398740050

 View more versions of this product

Catalog No. AC398740050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Ethyl 2-chloro-2-(hydroxyimino)acetate
96% min. (Argentometry)
C4H6ClNO3
5g
UXOLDCOJRAMLTQ-UTCJRWHESA-N
ethyl (2Z)-2-chloro-2-hydroxyiminoacetate
9576005
97%
14337-43-0
Glass bottle
MFCD00010209
ethyl 2-chloro-2-hydroxyimino acetate, 2-chloro-2-hydroxyiminoacetic acid ethyl ester, ethyl chlorooximinoacetate, ethyl chloro oximido acetate, ethyl chlorooximidoacetate, z-ethyl 2-chloro-2-hydroxyimino acetate, chloro oxime, ethyl 2z-2-chloro-2-hydroxyimino acetate, e-ethyl 2-chloro-2-hydroxyimino acetate, ethyl 2z-chloro hydroxyimino ethanoate
CCOC(=O)C(\Cl)=N\O
151.55
151.55

Chemical Identifiers

14337-43-0
151.55
UXOLDCOJRAMLTQ-UTCJRWHESA-N
9576005
CCOC(=O)C(\Cl)=N\O
C4H6ClNO3
MFCD00010209
ethyl 2-chloro-2-hydroxyimino acetate, 2-chloro-2-hydroxyiminoacetic acid ethyl ester, ethyl chlorooximinoacetate, ethyl chloro oximido acetate, ethyl chlorooximidoacetate, z-ethyl 2-chloro-2-hydroxyimino acetate, chloro oxime, ethyl 2z-2-chloro-2-hydroxyimino acetate, e-ethyl 2-chloro-2-hydroxyimino acetate, ethyl 2z-chloro hydroxyimino ethanoate
ethyl (2Z)-2-chloro-2-hydroxyiminoacetate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause an allergic skin reaction.
Causes serious eye damage.
Causes skin irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only