Learn More
Ethyl 2-chloro-2-(hydroxyimino)acetate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 398740250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ethyl 2-chloro-2-(hydroxyimino)acetate | |
96% min. (Argentometry) | |
C4H6ClNO3 | |
25g | |
UXOLDCOJRAMLTQ-UTCJRWHESA-N | |
ethyl (2Z)-2-chloro-2-hydroxyiminoacetate | |
9576005 | |
97% |
14337-43-0 | |
Glass bottle | |
MFCD00010209 | |
ethyl 2-chloro-2-hydroxyimino acetate, 2-chloro-2-hydroxyiminoacetic acid ethyl ester, ethyl chlorooximinoacetate, ethyl chloro oximido acetate, ethyl chlorooximidoacetate, z-ethyl 2-chloro-2-hydroxyimino acetate, chloro oxime, ethyl 2z-2-chloro-2-hydroxyimino acetate, e-ethyl 2-chloro-2-hydroxyimino acetate, ethyl 2z-chloro hydroxyimino ethanoate | |
CCOC(=O)C(\Cl)=N\O | |
151.55 | |
151.55 |
Chemical Identifiers
14337-43-0 | |
151.55 | |
UXOLDCOJRAMLTQ-UTCJRWHESA-N | |
9576005 | |
CCOC(=O)C(\Cl)=N\O |
C4H6ClNO3 | |
MFCD00010209 | |
ethyl 2-chloro-2-hydroxyimino acetate, 2-chloro-2-hydroxyiminoacetic acid ethyl ester, ethyl chlorooximinoacetate, ethyl chloro oximido acetate, ethyl chlorooximidoacetate, z-ethyl 2-chloro-2-hydroxyimino acetate, chloro oxime, ethyl 2z-2-chloro-2-hydroxyimino acetate, e-ethyl 2-chloro-2-hydroxyimino acetate, ethyl 2z-chloro hydroxyimino ethanoate | |
ethyl (2Z)-2-chloro-2-hydroxyiminoacetate |
Safety and Handling
GHS H Statement:
May cause an allergic skin reaction.
Causes serious eye damage.
Causes skin irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specificatio
GHS Signal Word: Danger
RUO â Research Use Only