Learn More
Ethyl 2-mercaptoacetate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 118675000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl 2-mercaptoacetate | |
623-51-8 | |
Colorless | |
54°C (12.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4560 to 1.459 | |
MFCD00004874 | |
03,255 | |
ethyl thioglycolate, ethyl mercaptoacetate, ethyl 2-mercaptoacetate, mercaptoacetic acid ethyl ester, ethyl thioglycollate, acetic acid, mercapto-, ethyl ester, thioglycolic acid ethyl ester, ethyl mercaptoacetic acid, usaf ek-2070, acetic acid, 2-mercapto-, ethyl ester | |
PVBRSNZAOAJRKO-UHFFFAOYSA-N | |
ethyl 2-sulfanylacetate | |
12185 | |
120.17 | |
Liquid |
99% | |
-80°C | |
1.0900g/mL | |
47°C | |
98.5% min. (GC) | |
C4H8O2S | |
HSCH2CO2C2H5 | |
500 g | |
1.09 | |
Solubility in water: insoluble. Other solubilities: soluble in ethanol,acetone | |
CCOC(=O)CS | |
120.17 | |
1.2 mPa.s (20°C) | |
99% |
Chemical Identifiers
623-51-8 | |
120.17 | |
PVBRSNZAOAJRKO-UHFFFAOYSA-N | |
12185 | |
CCOC(=O)CS |
C4H8O2S | |
MFCD00004874 | |
ethyl thioglycolate, ethyl mercaptoacetate, ethyl 2-mercaptoacetate, mercaptoacetic acid ethyl ester, ethyl thioglycollate, acetic acid, mercapto-, ethyl ester, thioglycolic acid ethyl ester, ethyl mercaptoacetic acid, usaf ek-2070, acetic acid, 2-mercapto-, ethyl ester | |
ethyl 2-sulfanylacetate |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 210-800-3
RTECSNumber : AI6650000
TSCA : TSCA
RUO – Research Use Only