missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2-methylquinoline-6-carboxylate, 97%, Thermo Scientific Chemicals

$103.36 - $367.07

Chemical Identifiers

CAS 855763-77-8
Molecular Formula C13H13NO2
Molecular Weight (g/mol) 215.252
MDL Number MFCD09879697
InChI Key WLKKUWXLOSMSMH-UHFFFAOYSA-N
Synonym 2-methylquinoline-6-carboxylic acid ethyl ester, ethyl 2-methyl-6-quinolinecarboxylate, ethyl2-methylquinoline-6-carboxylate, pubchem20820, 2-methylquinoline-6-carboxylic acidethylester, 2-methyl-6-quinolinecarboxylic acid ethyl ester, 6-quinolinecarboxylicacid, 2-methyl-, ethyl ester
PubChem CID 29949719
IUPAC Name ethyl 2-methylquinoline-6-carboxylate
SMILES CCOC(=O)C1=CC2=C(C=C1)N=C(C=C2)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6321303
View Documents
Thermo Scientific Chemicals
H6321303
1 g
Each for $103.36
 
AAH6321306
View Documents
Thermo Scientific Chemicals
H6321306
5 g
Each for $367.07
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

855763-77-8
215.252
WLKKUWXLOSMSMH-UHFFFAOYSA-N
29949719
CCOC(=O)C1=CC2=C(C=C1)N=C(C=C2)C
C13H13NO2
MFCD09879697
2-methylquinoline-6-carboxylic acid ethyl ester, ethyl 2-methyl-6-quinolinecarboxylate, ethyl2-methylquinoline-6-carboxylate, pubchem20820, 2-methylquinoline-6-carboxylic acidethylester, 2-methyl-6-quinolinecarboxylic acid ethyl ester, 6-quinolinecarboxylicacid, 2-methyl-, ethyl ester
ethyl 2-methylquinoline-6-carboxylate

Specifications

855763-77-8
MFCD09879697
2-methylquinoline-6-carboxylic acid ethyl ester, ethyl 2-methyl-6-quinolinecarboxylate, ethyl2-methylquinoline-6-carboxylate, pubchem20820, 2-methylquinoline-6-carboxylic acidethylester, 2-methyl-6-quinolinecarboxylic acid ethyl ester, 6-quinolinecarboxylicacid, 2-methyl-, ethyl ester
CCOC(=O)C1=CC2=C(C=C1)N=C(C=C2)C
215.252
215.25
Solid
C13H13NO2
1 g
WLKKUWXLOSMSMH-UHFFFAOYSA-N
ethyl 2-methylquinoline-6-carboxylate
29949719
97%
Ethyl 2-methylquinoline-6-carboxylate
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only