missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2-oxocyclopentanecarboxylate, 97+%, Thermo Scientific Chemicals

$78.89 - $224.62

Chemical Identifiers

CAS 611-10-9
Molecular Formula C8H12O3
Molecular Weight (g/mol) 156.181
MDL Number MFCD00001412
InChI Key JHZPNBKZPAWCJD-UHFFFAOYSA-N
Synonym ethyl 2-oxocyclopentanecarboxylate, 2-carbethoxycyclopentanone, ethyl cyclopentanone-2-carboxylate, cyclopentanecarboxylic acid, 2-oxo-, ethyl ester, cyclopentanone-2-carboxylic acid ethyl ester, ethyl2-oxocyclopentanecarboxylate, 2-ethoxycarbonyl cyclopentanone, ethyl 2-cyclopentanonecarboxylate, .alpha.-carboethoxy cyclopentanone, ethyl 2-cyclopentanone-1-carboxylate
PubChem CID 69136
IUPAC Name ethyl 2-oxocyclopentane-1-carboxylate
SMILES CCOC(=O)C1CCCC1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0452814
View Documents
Thermo Scientific Chemicals
L0452814
25 g
Each for $78.89
 
AAL0452822
View Documents
Thermo Scientific Chemicals
L0452822
100 g
Each for $224.62
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

611-10-9
156.181
JHZPNBKZPAWCJD-UHFFFAOYSA-N
69136
CCOC(=O)C1CCCC1=O
C8H12O3
MFCD00001412
ethyl 2-oxocyclopentanecarboxylate, 2-carbethoxycyclopentanone, ethyl cyclopentanone-2-carboxylate, cyclopentanecarboxylic acid, 2-oxo-, ethyl ester, cyclopentanone-2-carboxylic acid ethyl ester, ethyl2-oxocyclopentanecarboxylate, 2-ethoxycarbonyl cyclopentanone, ethyl 2-cyclopentanonecarboxylate, .alpha.-carboethoxy cyclopentanone, ethyl 2-cyclopentanone-1-carboxylate
ethyl 2-oxocyclopentane-1-carboxylate

Specifications

611-10-9
225°C to 228°C
C8H12O3
MFCD00001412
387787
JHZPNBKZPAWCJD-UHFFFAOYSA-N
ethyl 2-oxocyclopentane-1-carboxylate
69136
≥97%
1.079
77°C (170°F)
1.452
25 g
ethyl 2-oxocyclopentanecarboxylate, 2-carbethoxycyclopentanone, ethyl cyclopentanone-2-carboxylate, cyclopentanecarboxylic acid, 2-oxo-, ethyl ester, cyclopentanone-2-carboxylic acid ethyl ester, ethyl2-oxocyclopentanecarboxylate, 2-ethoxycarbonyl cyclopentanone, ethyl 2-cyclopentanonecarboxylate, .alpha.-carboethoxy cyclopentanone, ethyl 2-cyclopentanone-1-carboxylate
CCOC(=O)C1CCCC1=O
156.181
156.18
Ethyl 2-oxocyclopentanecarboxylate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 210-253-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only