missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Piperidinecarboxylate 98.0+%, TCI America™

$113.71 - $354.17

Chemical Identifiers

CAS 15862-72-3
Molecular Formula C8H16NO2
Molecular Weight (g/mol) 158.22
MDL Number MFCD00005980
InChI Key SZIKRGHFZTYTIT-ZETCQYMHSA-O
Synonym ethyl pipecolinate, ethyl 2-piperidinecarboxylate, ethyl pipecolate, pipecolinic acid ethyl ester, pipecolic acid ethyl ester, ethylpiperidine-2-carboxylate, 2-piperidinecarboxylic acid ethyl ester, 2-piperidinecarboxylic acid, ethyl ester, ethylpipecolinate, pubchem7720
PubChem CID 27517
IUPAC Name (2S)-2-(ethoxycarbonyl)piperidin-1-ium
SMILES CCOC(=O)[C@@H]1CCCC[NH2+]1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
E08375G
View Documents
TCI America
E08375G
5 g N/A
Only null left
 
E083725G
View Documents
TCI America
E083725G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

15862-72-3
158.22
SZIKRGHFZTYTIT-ZETCQYMHSA-O
27517
CCOC(=O)[C@@H]1CCCC[NH2+]1
C8H16NO2
MFCD00005980
ethyl pipecolinate, ethyl 2-piperidinecarboxylate, ethyl pipecolate, pipecolinic acid ethyl ester, pipecolic acid ethyl ester, ethylpiperidine-2-carboxylate, 2-piperidinecarboxylic acid ethyl ester, 2-piperidinecarboxylic acid, ethyl ester, ethylpipecolinate, pubchem7720
(2S)-2-(ethoxycarbonyl)piperidin-1-ium

Specifications

15862-72-3
217°C
MFCD00005980
1993
SZIKRGHFZTYTIT-ZETCQYMHSA-O
(2S)-2-(ethoxycarbonyl)piperidin-1-ium
27517
≥98.0% (GC,T)
Ethyl 2-Piperidinecarboxylate
Yellow
C8H16NO2
5 g
ethyl pipecolinate, ethyl 2-piperidinecarboxylate, ethyl pipecolate, pipecolinic acid ethyl ester, pipecolic acid ethyl ester, ethylpiperidine-2-carboxylate, 2-piperidinecarboxylic acid ethyl ester, 2-piperidinecarboxylic acid, ethyl ester, ethylpipecolinate, pubchem7720
CCOC(=O)[C@@H]1CCCC[NH2+]1
158.22
157.21
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'