missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3,4-dihydroxybenzoate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1100130

 View more versions of this product

Catalog No. AAA1100130


Explore available promotions

Description

Description

The compound is a prolyl 4-hydroxylase inhibitor and can be used to protect the myocardium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The compound is a prolyl 4-hydroxylase inhibitor and can be used to protect the myocardium.

Solubility
Insoluble in water. Soluble in ethanol.

Notes
Keep container tightly closed in a cool, dry and well-ventilated place. Store away from oxidizing agents, Strong bases.
Specifications

Specifications

Ethyl 3,4-dihydroxybenzoate
132°C to 135°C
MFCD00002199
2097435
Insoluble in water. Soluble in ethanol.
CCOC(=O)C1=CC=C(O)C(O)=C1
182.18
182.18
3943-89-3
C9H10O4
250 g
ethyl protocatechuate, 3,4-dihydroxybenzoic acid ethyl ester, ethyl-3,4-dihydroxybenzoate, unii-4ygj96wtbg, benzoic acid, 3,4-dihydroxy-, ethyl ester, edhb, protocatechuic acid ethyl ester, 4ygj96wtbg, 3,4-dihydroxy-benzoic acid ethyl ester
KBPUBCVJHFXPOC-UHFFFAOYSA-N
ethyl 3,4-dihydroxybenzoate
77547
98%

Chemical Identifiers

3943-89-3
182.18
KBPUBCVJHFXPOC-UHFFFAOYSA-N
77547
CCOC(=O)C1=CC=C(O)C(O)=C1
C9H10O4
MFCD00002199
ethyl protocatechuate, 3,4-dihydroxybenzoic acid ethyl ester, ethyl-3,4-dihydroxybenzoate, unii-4ygj96wtbg, benzoic acid, 3,4-dihydroxy-, ethyl ester, edhb, protocatechuic acid ethyl ester, 4ygj96wtbg, 3,4-dihydroxy-benzoic acid ethyl ester
ethyl 3,4-dihydroxybenzoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 223-529-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only