missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 3-(4-methoxybenzoyl)propionate, 98%, Thermo Scientific Chemicals
$128.03 - $395.99
Chemical Identifiers
CAS | 15118-67-9 |
---|---|
Molecular Formula | C13H16O4 |
Molecular Weight (g/mol) | 236.267 |
MDL Number | MFCD00051784 |
InChI Key | FYUAOZFEVHSJTO-UHFFFAOYSA-N |
Synonym | ethyl 3-4-methoxybenzoyl propionate, ethyl 4-4-methoxyphenyl-4-oxobutanoate, ethyl 4-4-methoxyphenyl-4-oxobutyrate, acmc-1ciqg, ethyl 3-4-methoxybenzoylpropionate, ethyl 3-4-methoxybenzoyl propanoate, ethyl 3-4-methoxybenzoyl-propionate, ethyl 4-4-methoxyphenyl-4-oxobutanoate #, gamma-oxo-4-methoxybenzenebutyric acid ethyl ester, 4-4-methoxyphenyl-4-oxobutyric acid, ethyl ester |
PubChem CID | 585132 |
IUPAC Name | ethyl 4-(4-methoxyphenyl)-4-oxobutanoate |
SMILES | CCOC(=O)CCC(=O)C1=CC=C(C=C1)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0798906
|
Thermo Scientific Chemicals
L0798906 |
5 g |
Each for $128.03
|
|
|||||
AAL0798914
|
Thermo Scientific Chemicals
L0798914 |
25 g |
Each for $395.99
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15118-67-9 | |
236.267 | |
FYUAOZFEVHSJTO-UHFFFAOYSA-N | |
585132 | |
CCOC(=O)CCC(=O)C1=CC=C(C=C1)OC |
C13H16O4 | |
MFCD00051784 | |
ethyl 3-4-methoxybenzoyl propionate, ethyl 4-4-methoxyphenyl-4-oxobutanoate, ethyl 4-4-methoxyphenyl-4-oxobutyrate, acmc-1ciqg, ethyl 3-4-methoxybenzoylpropionate, ethyl 3-4-methoxybenzoyl propanoate, ethyl 3-4-methoxybenzoyl-propionate, ethyl 4-4-methoxyphenyl-4-oxobutanoate #, gamma-oxo-4-methoxybenzenebutyric acid ethyl ester, 4-4-methoxyphenyl-4-oxobutyric acid, ethyl ester | |
ethyl 4-(4-methoxyphenyl)-4-oxobutanoate |
Specifications
15118-67-9 | |
C13H16O4 | |
5 g | |
ethyl 3-4-methoxybenzoyl propionate, ethyl 4-4-methoxyphenyl-4-oxobutanoate, ethyl 4-4-methoxyphenyl-4-oxobutyrate, acmc-1ciqg, ethyl 3-4-methoxybenzoylpropionate, ethyl 3-4-methoxybenzoyl propanoate, ethyl 3-4-methoxybenzoyl-propionate, ethyl 4-4-methoxyphenyl-4-oxobutanoate #, gamma-oxo-4-methoxybenzenebutyric acid ethyl ester, 4-4-methoxyphenyl-4-oxobutyric acid, ethyl ester | |
CCOC(=O)CCC(=O)C1=CC=C(C=C1)OC | |
236.267 | |
236.27 | |
Ethyl 3-(4-methoxybenzoyl)propionate |
49°C to 53°C | |
MFCD00051784 | |
2120509 | |
FYUAOZFEVHSJTO-UHFFFAOYSA-N | |
ethyl 4-(4-methoxyphenyl)-4-oxobutanoate | |
585132 | |
98% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only