missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate, 96%, Thermo Scientific Chemicals

$675.68 - $675.68

Chemical Identifiers

CAS 128073-16-5
Molecular Formula C9H7ClF3NO2
Molecular Weight (g/mol) 253.605
MDL Number MFCD06656414
InChI Key COMQYNZHBCNPNW-UHFFFAOYSA-N
Synonym ethyl 3-chloro-5-trifluoromethyl picolinate, ethyl 3-chloro-5-trifluoromethyl pyridine-2-carboxylate, ethyl 3-chloro-5-trifluoromethyl-2-pyridinecarboxylate, 2-pyridinecarboxylic acid, 3-chloro-5-trifluoromethyl-, ethyl ester, 3-chloro-5-trifluoromethylpyridine-2-carboxylic acid ethyl ester, ethyl 5-trifluoromethyl-3-chloropyridine-2-carboxylate, ethyl 3-chloro-5-trifluoromethyl picolite, ethyl 3-chloro-5trifluoromethyl-2-pyridinecarboxylate
PubChem CID 22013558
IUPAC Name ethyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate
SMILES CCOC(=O)C1=C(C=C(C=N1)C(F)(F)F)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3396709
View Documents
Thermo Scientific Chemicals
H3396709
10 g
Each for $675.68
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

128073-16-5
253.605
COMQYNZHBCNPNW-UHFFFAOYSA-N
22013558
CCOC(=O)C1=C(C=C(C=N1)C(F)(F)F)Cl
C9H7ClF3NO2
MFCD06656414
ethyl 3-chloro-5-trifluoromethyl picolinate, ethyl 3-chloro-5-trifluoromethyl pyridine-2-carboxylate, ethyl 3-chloro-5-trifluoromethyl-2-pyridinecarboxylate, 2-pyridinecarboxylic acid, 3-chloro-5-trifluoromethyl-, ethyl ester, 3-chloro-5-trifluoromethylpyridine-2-carboxylic acid ethyl ester, ethyl 5-trifluoromethyl-3-chloropyridine-2-carboxylate, ethyl 3-chloro-5-trifluoromethyl picolite, ethyl 3-chloro-5trifluoromethyl-2-pyridinecarboxylate
ethyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate

Specifications

128073-16-5
C9H7ClF3NO2
10 g
COMQYNZHBCNPNW-UHFFFAOYSA-N
ethyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate
22013558
96%
64°C to 66°C (0.008 mmHg)
MFCD06656414
ethyl 3-chloro-5-trifluoromethyl picolinate, ethyl 3-chloro-5-trifluoromethyl pyridine-2-carboxylate, ethyl 3-chloro-5-trifluoromethyl-2-pyridinecarboxylate, 2-pyridinecarboxylic acid, 3-chloro-5-trifluoromethyl-, ethyl ester, 3-chloro-5-trifluoromethylpyridine-2-carboxylic acid ethyl ester, ethyl 5-trifluoromethyl-3-chloropyridine-2-carboxylate, ethyl 3-chloro-5-trifluoromethyl picolite, ethyl 3-chloro-5trifluoromethyl-2-pyridinecarboxylate
CCOC(=O)C1=C(C=C(C=N1)C(F)(F)F)Cl
253.605
253.6
Ethyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only