missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(Chloromethyl)benzoate 96.0+%, TCI America™

Supplier:  TCI America E06615G

 View more versions of this product

Catalog No. E06615G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethyl 3-(Chloromethyl)benzoate
169°C
5 g
3-(Chloromethyl)benzoic Acid Ethyl Ester
CCOC(=O)C1=CC(=CC=C1)CCl
198.646
198.65
Liquid
54589-54-7
C10H11ClO2
3265
ZJNVMXXFCNKXLT-UHFFFAOYSA-N
ethyl 3-(chloromethyl)benzoate
14812190
≥96.0% (GC)

Chemical Identifiers

54589-54-7
198.646
3-(Chloromethyl)benzoic Acid Ethyl Ester
ethyl 3-(chloromethyl)benzoate
C10H11ClO2
ZJNVMXXFCNKXLT-UHFFFAOYSA-N
14812190
CCOC(=O)C1=CC(=CC=C1)CCl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'

Promotions

Promotions