missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-cyano-3-phenylpyruvate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2695106

 View more versions of this product

Catalog No. AAH2695106


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl 3-cyano-3-phenylpyruvate
125°C to 129°C
MFCD00019801
ethyl 3-cyano-3-phenylpyruvate, ethyl phenylcyanopyruvate, cyanophenylpyruvic acid ethyl ester, ethyl cyanophenylpyruvate, ethyl alpha-cyanophenylpyruvate, pyruvic acid, cyanophenyl-, ethyl ester, ethyl .alpha.-cyanophenylpyruvate, wln: ncr x1v1uno2, 4-10-00-03440 beilstein handbook reference, benzenepropanoic acid, .beta.-cyano-.alpha.-oxo-, ethyl ester
CCOC(=O)C(=O)C(C#N)C1=CC=CC=C1
217.22
217.23
6362-63-6
C12H11NO3
5 g
YWNRATDDUSMBPR-UHFFFAOYNA-N
ethyl 3-cyano-2-oxo-3-phenylpropanoate
22845
97%

Chemical Identifiers

6362-63-6
217.22
YWNRATDDUSMBPR-UHFFFAOYNA-N
22845
CCOC(=O)C(=O)C(C#N)C1=CC=CC=C1
C12H11NO3
MFCD00019801
ethyl 3-cyano-3-phenylpyruvate, ethyl phenylcyanopyruvate, cyanophenylpyruvic acid ethyl ester, ethyl cyanophenylpyruvate, ethyl alpha-cyanophenylpyruvate, pyruvic acid, cyanophenyl-, ethyl ester, ethyl .alpha.-cyanophenylpyruvate, wln: ncr x1v1uno2, 4-10-00-03440 beilstein handbook reference, benzenepropanoic acid, .beta.-cyano-.alpha.-oxo-, ethyl ester
ethyl 3-cyano-2-oxo-3-phenylpropanoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 228-844-7

RTECSNumber : UZ0875000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only