missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-(dimethylamino)acrylate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2429130

 View more versions of this product

Catalog No. AAB2429130


Explore available promotions

Description

Description

Ethyl 3-(dimethylamino)acrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-(dimethylamino)acrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in water 90 g/L (25°C).

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Specifications

Specifications

Ethyl 3-(dimethylamino)acrylate
17°C to 18°C
C7H13NO2
1.512
250 g
Soluble in water 90g/L (25°C).
CCOC(=O)C=CN(C)C
143.186
143.19
924-99-2
118°C to 121°C (10 mmHg)
(CH3)2NCH=CHCO2CH2CH3
MFCD00144269
ethyl 3-dimethylamino acrylate, e-ethyl 3-dimethylamino acrylate, ethyl 3-n,n-dimethylamino acrylate, ethyl n,n-dimethylaminoacrylate, ethyl trans-3-dimethylaminoacrylate, ethyl 2e-3-dimethylamino prop-2-enoate, ethyl e-3-dimethylamino prop-2-enoate, ethyl 3-dimethylamino prop-2-enoate, acrylic acid, 3-dimethylamino-, ethyl ester, e, 2-propenoic acid, 3-dimethylamino-, ethyl ester, 2e
MVUMJYQUKKUOHO-AATRIKPKSA-N
ethyl (E)-3-(dimethylamino)prop-2-enoate
5369162
99%

Chemical Identifiers

924-99-2
143.186
MVUMJYQUKKUOHO-AATRIKPKSA-N
5369162
CCOC(=O)C=CN(C)C
C7H13NO2
MFCD00144269
ethyl 3-dimethylamino acrylate, e-ethyl 3-dimethylamino acrylate, ethyl 3-n,n-dimethylamino acrylate, ethyl n,n-dimethylaminoacrylate, ethyl trans-3-dimethylaminoacrylate, ethyl 2e-3-dimethylamino prop-2-enoate, ethyl e-3-dimethylamino prop-2-enoate, ethyl 3-dimethylamino prop-2-enoate, acrylic acid, 3-dimethylamino-, ethyl ester, e, 2-propenoic acid, 3-dimethylamino-, ethyl ester, 2e
ethyl (E)-3-(dimethylamino)prop-2-enoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H317
May cause an allergic skin reaction.

P261-P272-P280g-P302+P352-P333+P313-P363-P501c

H317

EINECSNumber : 402-650-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only