missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-(Dimethylamino)acrylate 98.0+%, TCI America™

Supplier:  TCI America D283625G

 View more versions of this product

Catalog No. D283625G


Explore available promotions

Description

Description

Specifications

Specifications

Ethyl 3-(Dimethylamino)acrylate
19°C
136°C
MFCD00144269
ethyl 3-dimethylamino acrylate, e-ethyl 3-dimethylamino acrylate, ethyl 3-n,n-dimethylamino acrylate, ethyl n,n-dimethylaminoacrylate, ethyl trans-3-dimethylaminoacrylate, ethyl 2e-3-dimethylamino prop-2-enoate, ethyl e-3-dimethylamino prop-2-enoate, ethyl 3-dimethylamino prop-2-enoate, acrylic acid, 3-dimethylamino-, ethyl ester, e, 2-propenoic acid, 3-dimethylamino-, ethyl ester, 2e
CCOC(=O)C=CN(C)C
143.186
143.19
Liquid
924-99-2
Yellow
C7H13NO2
25 g
MVUMJYQUKKUOHO-AATRIKPKSA-N
ethyl (E)-3-(dimethylamino)prop-2-enoate
5369162
≥98.0% (GC,T)

Chemical Identifiers

924-99-2
143.186
MVUMJYQUKKUOHO-AATRIKPKSA-N
5369162
CCOC(=O)C=CN(C)C
C7H13NO2
MFCD00144269
ethyl 3-dimethylamino acrylate, e-ethyl 3-dimethylamino acrylate, ethyl 3-n,n-dimethylamino acrylate, ethyl n,n-dimethylaminoacrylate, ethyl trans-3-dimethylaminoacrylate, ethyl 2e-3-dimethylamino prop-2-enoate, ethyl e-3-dimethylamino prop-2-enoate, ethyl 3-dimethylamino prop-2-enoate, acrylic acid, 3-dimethylamino-, ethyl ester, e, 2-propenoic acid, 3-dimethylamino-, ethyl ester, 2e
ethyl (E)-3-(dimethylamino)prop-2-enoate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3661

TSCA : No

SDS
Documents

Documents

Promotions

Promotions