Learn More
Ethyl 3-dimethylaminopropionate, 97%
CAS: 20120-21-2 | C7H15NO2 | 145.20 g/mol
$210.07 - $210.07
Chemical Identifiers
| CAS | 20120-21-2 |
|---|---|
| Molecular Formula | C7H15NO2 |
| Molecular Weight (g/mol) | 145.20 |
| MDL Number | MFCD00015166 |
| InChI Key | IZMJWGJGVGRWBW-UHFFFAOYSA-N |
| Synonym | ethyl 3-dimethylamino propanoate, ethyl 3-dimethylaminopropionate, ethyl 3-dimethylamino propionate, n,n-dimethyl-beta-alanine ethyl ester, ethyl 3-n,n-dimethylamino propionate, ethyl n,n-dimethyl-beta-alaninate, ethyl beta-dimethylamino propionate, beta-alanine, n,n-dimethyl-, ethyl ester, ethyl 3-dimethyl-aminopropionate, ethyl-3-n,n-dimethylamino propionate |
| PubChem CID | 88372 |
| IUPAC Name | ethyl 3-(dimethylamino)propanoate |
| SMILES | CCOC(=O)CCN(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0154214
|
Thermo Scientific Chemicals
L0154214 |
25 g |
Each for $210.07
|
|
|||||
Description
Ethyl 3-dimethylaminopropionate is used in the preparation of [3-(dimethylamino)propionyl]guanidine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 3-dimethylaminopropionate is used in the preparation of [3-(dimethylamino)propionyl]guanidine.
Solubility
Immiscible with water.
Notes
Incompatible with oxidizing agents.
Chemical Identifiers
| 20120-21-2 | |
| 145.20 | |
| IZMJWGJGVGRWBW-UHFFFAOYSA-N | |
| 88372 | |
| CCOC(=O)CCN(C)C |
| C7H15NO2 | |
| MFCD00015166 | |
| ethyl 3-dimethylamino propanoate, ethyl 3-dimethylaminopropionate, ethyl 3-dimethylamino propionate, n,n-dimethyl-beta-alanine ethyl ester, ethyl 3-n,n-dimethylamino propionate, ethyl n,n-dimethyl-beta-alaninate, ethyl beta-dimethylamino propionate, beta-alanine, n,n-dimethyl-, ethyl ester, ethyl 3-dimethyl-aminopropionate, ethyl-3-n,n-dimethylamino propionate | |
| ethyl 3-(dimethylamino)propanoate |
Specifications
| 20120-21-2 | |
| 56°C to 57°C (12 mmHg) | |
| Amine-like | |
| 1.419 | |
| 25 g | |
| 1702477 | |
| Immiscible with water. | |
| CCOC(=O)CCN(C)C | |
| 145.20 | |
| 145.2 | |
| Ethyl 3-dimethylaminopropionate |
| 0.915 | |
| 58°C (136°F) | |
| C7H15NO2 | |
| MFCD00015166 | |
| UN1993 | |
| ethyl 3-dimethylamino propanoate, ethyl 3-dimethylaminopropionate, ethyl 3-dimethylamino propionate, n,n-dimethyl-beta-alanine ethyl ester, ethyl 3-n,n-dimethylamino propionate, ethyl n,n-dimethyl-beta-alaninate, ethyl beta-dimethylamino propionate, beta-alanine, n,n-dimethyl-, ethyl ester, ethyl 3-dimethyl-aminopropionate, ethyl-3-n,n-dimethylamino propionate | |
| IZMJWGJGVGRWBW-UHFFFAOYSA-N | |
| ethyl 3-(dimethylamino)propanoate | |
| 88372 | |
| 97% |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 243-524-7
RTECSNumber : AY5992700
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only