missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-Methylvalerate 98.0+%, TCI America™
Supplier: TCI America E08035G
Specifications
| Ethyl 3-Methylvalerate | |
| Colorless | |
| C8H16O2 | |
| 5 g | |
| ethyl 3-methylvalerate, pentanoic acid, 3-methyl-, ethyl ester, ethyl beta-methylvalerate, valeric acid, 3-methyl-, ethyl ester, 3-methylvaleric acid ethyl ester, 3-methylpentanoic acid, ethyl ester, fema no. 3679, ethyl 3-methyl valerate, ethyl 3-methyl pentanoate | |
| CCC(C)CC(=O)OCC | |
| 144.214 | |
| 144.21 | |
| Liquid |
| 5870-68-8 | |
| 158°C | |
| MFCD00041390 | |
| 3272 | |
| TXAWGHYFBQBVNK-UHFFFAOYSA-N | |
| ethyl 3-methylpentanoate | |
| 560255 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 5870-68-8 | |
| 144.214 | |
| TXAWGHYFBQBVNK-UHFFFAOYSA-N | |
| 560255 | |
| CCC(C)CC(=O)OCC |
| C8H16O2 | |
| MFCD00041390 | |
| ethyl 3-methylvalerate, pentanoic acid, 3-methyl-, ethyl ester, ethyl beta-methylvalerate, valeric acid, 3-methyl-, ethyl ester, 3-methylvaleric acid ethyl ester, 3-methylpentanoic acid, ethyl ester, fema no. 3679, ethyl 3-methyl valerate, ethyl 3-methyl pentanoate | |
| ethyl 3-methylpentanoate |
Safety and Handling
EINECSNumber : (2)-0782
TSCA : No