missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-Oxo-3-(2-thienyl)propionate 98.0+%, TCI America™
$72.70 - $214.37
Chemical Identifiers
| CAS | 13669-10-8 |
|---|---|
| Molecular Formula | C9H10O3S |
| Molecular Weight (g/mol) | 198.236 |
| MDL Number | MFCD00542649 |
| InChI Key | VKSDKUXHVLZDHO-UHFFFAOYSA-N |
| Synonym | 3-Oxo-3-(2-thienyl)propionic Acid Ethyl Ester |
| PubChem CID | 255159 |
| IUPAC Name | ethyl 3-oxo-3-thiophen-2-ylpropanoate |
| SMILES | CCOC(=O)CC(=O)C1=CC=CS1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
E1073200MG
|
TCI America
E1073200MG |
200 mg |
Each for $72.70
|
|
|||||
|
E10731G
|
TCI America
E10731G |
1 g |
Each for $214.37
|
|
|||||
Chemical Identifiers
| 13669-10-8 | |
| 198.236 | |
| VKSDKUXHVLZDHO-UHFFFAOYSA-N | |
| 255159 | |
| CCOC(=O)CC(=O)C1=CC=CS1 |
| C9H10O3S | |
| MFCD00542649 | |
| 3-Oxo-3-(2-thienyl)propionic Acid Ethyl Ester | |
| ethyl 3-oxo-3-thiophen-2-ylpropanoate |
Specifications
| 13669-10-8 | |
| C9H10O3S | |
| 200 mg | |
| VKSDKUXHVLZDHO-UHFFFAOYSA-N | |
| ethyl 3-oxo-3-thiophen-2-ylpropanoate | |
| 255159 | |
| ≥98.0% (GC) | |
| Ethyl 3-Oxo-3-(2-thienyl)propionate |
| 130°C | |
| MFCD00542649 | |
| 3-Oxo-3-(2-thienyl)propionic Acid Ethyl Ester | |
| CCOC(=O)CC(=O)C1=CC=CS1 | |
| 198.236 | |
| 198.24 | |
| Liquid |
Safety and Handling
TSCA : No