missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 3-Oxo-3-(4-pyridyl)propionate 98.0+%, TCI America™
Supplier: TCI America E10325G
Specifications
Ethyl 3-Oxo-3-(4-pyridyl)propionate | |
57°C | |
169°C | |
MFCD00094021 | |
ethyl isonicotinoylacetate, ethyl 3-oxo-3-pyridin-4-yl propanoate, ethyl 3-oxo-3-4-pyridyl propionate, ethyl 3-4-pyridyl-3-oxopropionate, 3-oxo-3-pyridin-4-yl-propionic acid ethyl ester, ethyl 3-oxo-3-4-pyridinyl propanoate, ethylisonicotinoylacetate, ethyl-4-pyridoyl acetate, ethyl 3-oxo-3-4-pyridyl propanoate, 4-pyridinepropanoic acid, b-oxo-, ethyl ester | |
CCOC(=O)CC(=O)C1=CC=NC=C1 | |
193.20 | |
193.20 | |
Crystalline Powder |
26377-17-3 | |
White-Yellow | |
C10H11NO3 | |
5 g | |
PCJNYGPKMQQCPX-UHFFFAOYSA-N | |
ethyl 3-oxo-3-(pyridin-4-yl)propanoate | |
2735202 | |
≥98.0% (GC,T) |
Chemical Identifiers
26377-17-3 | |
193.20 | |
PCJNYGPKMQQCPX-UHFFFAOYSA-N | |
2735202 | |
CCOC(=O)CC(=O)C1=CC=NC=C1 |
C10H11NO3 | |
MFCD00094021 | |
ethyl isonicotinoylacetate, ethyl 3-oxo-3-pyridin-4-yl propanoate, ethyl 3-oxo-3-4-pyridyl propionate, ethyl 3-4-pyridyl-3-oxopropionate, 3-oxo-3-pyridin-4-yl-propionic acid ethyl ester, ethyl 3-oxo-3-4-pyridinyl propanoate, ethylisonicotinoylacetate, ethyl-4-pyridoyl acetate, ethyl 3-oxo-3-4-pyridyl propanoate, 4-pyridinepropanoic acid, b-oxo-, ethyl ester | |
ethyl 3-oxo-3-(pyridin-4-yl)propanoate |
Safety and Handling
TSCA : No