missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-(trimethylsilyl)propiolate, 98%, Thermo Scientific Chemicals

$111.77 - $365.23

Chemical Identifiers

CAS 16205-84-8
Molecular Formula C8H14O2Si
Molecular Weight (g/mol) 170.283
MDL Number MFCD00074886
InChI Key QRBKPHYAIRLCLK-UHFFFAOYSA-N
Synonym ethyl 3-trimethylsilyl propiolate, ethyl 3-trimethylsilyl propynoate, ethyl 3-trimethylsilyl prop-2-ynoate, 2-propynoic acid, 3-trimethylsilyl-, ethyl ester, ethyl 3-trimethylsilyl-2-propynoate, acmc-1bvr4, ethyl 3-trimethylsily propynoate, ethyl3-trimethylsilyl propiolate, ethyl 3-trimethylsilyl propalate, trimethylsilylpropiolic acid ethyl ester
PubChem CID 568601
IUPAC Name ethyl 3-trimethylsilylprop-2-ynoate
SMILES CCOC(=O)C#C[Si](C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5351706
View Documents
Thermo Scientific Chemicals
H5351706
5 g
Each for $111.77
 
AAH5351714
View Documents
Thermo Scientific Chemicals
H5351714
25 g
Each for $365.23
 
Description

Description

Ethyl 3-(trimethylsilyl)propiolate was used in the preparation of 4-ethoxycarbonyl-3-(trimethylsilyl)furan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-(trimethylsilyl)propiolate was used in the preparation of 4-ethoxycarbonyl-3-(trimethylsilyl)furan.

Solubility
Sparingly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

16205-84-8
170.283
QRBKPHYAIRLCLK-UHFFFAOYSA-N
568601
CCOC(=O)C#C[Si](C)(C)C
C8H14O2Si
MFCD00074886
ethyl 3-trimethylsilyl propiolate, ethyl 3-trimethylsilyl propynoate, ethyl 3-trimethylsilyl prop-2-ynoate, 2-propynoic acid, 3-trimethylsilyl-, ethyl ester, ethyl 3-trimethylsilyl-2-propynoate, acmc-1bvr4, ethyl 3-trimethylsily propynoate, ethyl3-trimethylsilyl propiolate, ethyl 3-trimethylsilyl propalate, trimethylsilylpropiolic acid ethyl ester
ethyl 3-trimethylsilylprop-2-ynoate

Specifications

92°C to 96°C (28mmHg)
5 g
(CH3)3SiC≡CCO2CH2CH3
1767850
ethyl 3-trimethylsilyl propiolate, ethyl 3-trimethylsilyl propynoate, ethyl 3-trimethylsilyl prop-2-ynoate, 2-propynoic acid, 3-trimethylsilyl-, ethyl ester, ethyl 3-trimethylsilyl-2-propynoate, acmc-1bvr4, ethyl 3-trimethylsily propynoate, ethyl3-trimethylsilyl propiolate, ethyl 3-trimethylsilyl propalate, trimethylsilylpropiolic acid ethyl ester
QRBKPHYAIRLCLK-UHFFFAOYSA-N
ethyl 3-trimethylsilylprop-2-ynoate
568601
98%
0.897
16205-84-8
C8H14O2Si
MFCD00074886
71°C (160°F)
Sparingly soluble in water.
CCOC(=O)C#C[Si](C)(C)C
170.283
170.28
1.441
Ethyl 3-(trimethylsilyl)propiolate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P370+P378q-P501c

H227-H319-H335

DOTInformation : Hazard Class: 9; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only