missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 4-aminobenzoate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 150785000

 View more versions of this product

Catalog No. AC150785000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl 4-aminobenzoate
89.0°C to 92.0°C
172°C (17.0 hPa)
Authentic
Plastic bottle
H2NC6H4CO2C2H5
500 g
15,1088
Solubility in water: 1g/2500mL. Other solubilities: 1g/5mL alcohol,1g/2mL chloroform,1g/4mL ether,ca. 1g/40mL expressed almond oil or olive oil
CCOC(=O)C1=CC=C(N)C=C1
165.19
CHEBI:116735
98%
94-09-7
White
>100°C
97.5% min. (GC)
C9H11NO2
MFCD00007892
14,422
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
BLFLLBZGZJTVJG-UHFFFAOYSA-N
ethyl 4-aminobenzoate
2337
165.19
Crystalline Powder

Chemical Identifiers

94-09-7
165.19
BLFLLBZGZJTVJG-UHFFFAOYSA-N
2337
ethyl 4-aminobenzoate
C9H11NO2
MFCD00007892
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
CHEBI:116735
CCOC(=O)C1=CC=C(N)C=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 202-303-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only