Learn More
Ethyl 4-bromo-3-ethoxy-2-butenoate, tech. 90%, stab. with sodium carbonate
CAS: 1116-50-3 | C8H13BrO3 | 237.09 g/mol
$192.42 - $192.42
Chemical Identifiers
| CAS | 1116-50-3 |
|---|---|
| Molecular Formula | C8H13BrO3 |
| Molecular Weight (g/mol) | 237.09 |
| MDL Number | MFCD00051757 |
| InChI Key | FYYMWILFRUUBCX-UHFFFAOYSA-N |
| Synonym | ethyl 4-bromo-3-ethoxybut-2-enoate, ethyl 4-bromo-3-ethoxy-2-butenoate, ethyl 2e-4-bromo-3-ethoxybut-2-enoate, ethyl e-4-bromo-3-ethoxybut-2-enoate |
| PubChem CID | 6376123 |
| SMILES | CCOC(=O)C=C(CBr)OCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0684906
|
Thermo Scientific Chemicals
L0684906 |
5 g |
N/A
|
|
|||||
Description
Ethyl 4-bromo-3-ethoxy-2-butenoate is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 4-bromo-3-ethoxy-2-butenoate is used as pharmaceutical intermediate.
Solubility
Not miscible in water.
Notes
Moisture and light sensitive. Incompatible with oxidizing agents, bases and heat.
Chemical Identifiers
| 1116-50-3 | |
| 237.09 | |
| FYYMWILFRUUBCX-UHFFFAOYSA-N | |
| 6376123 |
| C8H13BrO3 | |
| MFCD00051757 | |
| ethyl 4-bromo-3-ethoxybut-2-enoate, ethyl 4-bromo-3-ethoxy-2-butenoate, ethyl 2e-4-bromo-3-ethoxybut-2-enoate, ethyl e-4-bromo-3-ethoxybut-2-enoate | |
| CCOC(=O)C=C(CBr)OCC |
Specifications
| 1116-50-3 | |
| Pungent | |
| 1.496 | |
| 5 g | |
| 2042896 | |
| ethyl 4-bromo-3-ethoxybut-2-enoate, ethyl 4-bromo-3-ethoxy-2-butenoate, ethyl 2e-4-bromo-3-ethoxybut-2-enoate, ethyl e-4-bromo-3-ethoxybut-2-enoate | |
| FYYMWILFRUUBCX-UHFFFAOYSA-N | |
| ethyl (E)-4-bromo-3-ethoxybut-2-enoate | |
| 6376123 | |
| 90% | |
| Ethyl 4-bromo-3-ethoxy-2-butenoate, stabilized with sodium carbonate |
| 88°C to 90°C (1 mmHg) | |
| C8H13BrO3 | |
| MFCD00051757 | |
| UN3265 | |
| Light and moisture sensitive | |
| Not miscible in water. | |
| CCOC(=O)C=C(CBr)OCC | |
| 237.09 | |
| 237.1 | |
| Technical |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only