missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 4-bromo-5-(trifluoromethyl)-1H-pyrazole-3-carboxylate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3406503

 View more versions of this product

Catalog No. AAH3406503


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl 4-bromo-5-(trifluoromethyl)-1H-pyrazole-3-carboxylate
131°C to 133°C
MFCD13193102
ethyl 4-bromo-5-trifluoromethyl-1h-pyrazole-3-carboxylate, ethylbromotrifluoromethylpyrazolecarboxylate, ethyl 4-bromo-3-trifluoromethyl-1h-pyrazole-5-carboxylate
CCOC(=O)C1=NNC(=C1Br)C(F)(F)F
287.036
287
1240278-21-0
C7H6BrF3N2O2
1 g
OVIHBAGNAYIHMQ-UHFFFAOYSA-N
ethyl 4-bromo-5-(trifluoromethyl)-1H-pyrazole-3-carboxylate
45588074
97%

Chemical Identifiers

1240278-21-0
287.036
OVIHBAGNAYIHMQ-UHFFFAOYSA-N
45588074
CCOC(=O)C1=NNC(=C1Br)C(F)(F)F
C7H6BrF3N2O2
MFCD13193102
ethyl 4-bromo-5-trifluoromethyl-1h-pyrazole-3-carboxylate, ethylbromotrifluoromethylpyrazolecarboxylate, ethyl 4-bromo-3-trifluoromethyl-1h-pyrazole-5-carboxylate
ethyl 4-bromo-5-(trifluoromethyl)-1H-pyrazole-3-carboxylate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only