missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 4-dimethylaminobenzoate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1288814

 View more versions of this product

Catalog No. AAA1288814


Explore available promotions

Description

Description

Ethyl 4-dimethylaminobenzoate is employed as a photoinititator in cell encapsulation applications. It is also useful for organic synthesis. Further, it is used in UV-curing coatings and inks for the initiation of photo polymerization of unsaturated prepolymers. In addition to this, it acts as a active pharmaceutical ingredient intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-dimethylaminobenzoate is employed as a photoinititator in cell encapsulation applications. It is also useful for organic synthesis. Further, it is used in UV-curing coatings and inks for the initiation of photo polymerization of unsaturated prepolymers. In addition to this, it acts as a active pharmaceutical ingredient intermediate.

Solubility
Insoluble in water.

Notes
Incompatible with reducing agents, oxidizing agents, bases and acids.
Specifications

Specifications

Ethyl 4-dimethylaminobenzoate
63°C to 65°C
190°C to 191°C (14 mmHg)
MFCD00009115
2210233
Insoluble in water.
CCOC(=O)C1=CC=C(C=C1)N(C)C
193.246
CHEBI:52073
99%
10287-53-3
1.06
C11H15NO2
25 g
ethyl 4-dimethylamino benzoate, parbenate, benzoic acid, 4-dimethylamino-, ethyl ester, kayacure epa, ethyl-p-dimethylaminobenzoate, ethyl-4-dimethylaminobenzoate, n,n-dimethylbenzocaine, speedcure edb, ethyl p-dimethylaminobenzoate, unii-829s8d3y0x
FZUGPQWGEGAKET-UHFFFAOYSA-N
ethyl 4-(dimethylamino)benzoate
25127
193.25

Chemical Identifiers

10287-53-3
193.246
FZUGPQWGEGAKET-UHFFFAOYSA-N
25127
ethyl 4-(dimethylamino)benzoate
C11H15NO2
MFCD00009115
ethyl 4-dimethylamino benzoate, parbenate, benzoic acid, 4-dimethylamino-, ethyl ester, kayacure epa, ethyl-p-dimethylaminobenzoate, ethyl-4-dimethylaminobenzoate, n,n-dimethylbenzocaine, speedcure edb, ethyl p-dimethylaminobenzoate, unii-829s8d3y0x
CHEBI:52073
CCOC(=O)C1=CC=C(C=C1)N(C)C
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 233-634-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only