missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 4-Hydroxy-3-methoxycinnamate 97.0+%, TCI America™

Supplier:  TCI America E07395G

 View more versions of this product

Catalog No. E07395G


Explore available promotions

Description

Description

Specifications

Specifications

Ethyl 4-Hydroxy-3-methoxycinnamate
57°C
185°C
MFCD00009190
ethyl ferulate, ferulic acid ethyl ester, ethyl 3-4-hydroxy-3-methoxyphenyl acrylate, unii-5b8915uelw, ethyl 4'-hydroxy-3'-methoxycinnamate, e-ethyl 3-4-hydroxy-3-methoxyphenyl acrylate, ethyl 2e-3-4-hydroxy-3-methoxyphenyl prop-2-enoate, ethyl 4-hydroxy-3-methoxycinnamate, e-3-4-hydroxy-3-methoxy-phenyl-acrylic acid ethyl ester, ethyl 4-hydroxy-3-methoxycinnamate ferulic acid ethyl ester
CCOC(=O)C=CC1=CC(=C(C=C1)O)OC
222.24
222.24
Crystalline Powder
4046-02-0
Yellow
C12H14O4
5 g
ATJVZXXHKSYELS-FNORWQNLSA-N
ethyl (E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoate
736681
≥97.0% (GC)

Chemical Identifiers

4046-02-0
222.24
ATJVZXXHKSYELS-FNORWQNLSA-N
736681
CCOC(=O)C=CC1=CC(=C(C=C1)O)OC
C12H14O4
MFCD00009190
ethyl ferulate, ferulic acid ethyl ester, ethyl 3-4-hydroxy-3-methoxyphenyl acrylate, unii-5b8915uelw, ethyl 4'-hydroxy-3'-methoxycinnamate, e-ethyl 3-4-hydroxy-3-methoxyphenyl acrylate, ethyl 2e-3-4-hydroxy-3-methoxyphenyl prop-2-enoate, ethyl 4-hydroxy-3-methoxycinnamate, e-3-4-hydroxy-3-methoxy-phenyl-acrylic acid ethyl ester, ethyl 4-hydroxy-3-methoxycinnamate ferulic acid ethyl ester
ethyl (E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoate
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions